Skip to Content
Merck
All Photos(1)

Key Documents

ABT1384

Sigma-Aldrich

Anti-Neurocan Antibody, N-terminal

from rabbit

Synonym(s):

Neurocan core protein, 245 kDa early postnatal core glycoprotein, Chondroitin sulfate proteoglycan 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable

isotype

IgG

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

rat ... Ncan(58982)

General description

Neurocan core protein (UniProt: P55067; also known as 245 kDa early postnatal core glycoprotein, Chondroitin sulfate proteoglycan 3) is encoded by the Ncan (also known as Cspg3) gene in rat. Neurocan is a secreted protein of the aggrecan/versican proteoglycan family that is found in early postnatal and adult brain. During early postnatal development neurocan accounts for at least 20% of the protein of the soluble brain proteoglycans. Neurocan mostly contains chondroitin sulfate, but also has N-linked and O-linked oligosaccharides. It is shown to be involved in the modulation of cell adhesion and migration and regulates neurite outgrowth. Neurocan interact with several cell surface molecules, such as N-CAM, L1/Ng-CAM, etc., and modulates the cell-binding and neurite outgrowth promoting activates of these molecules. Neurocan is synthesized with a 22 aa signal peptide that is subsequently cleaved to produce the mature form. Neurocan contains one C-type lectin domain, two EGF-like domains, one Ig-like V type domain, two link domains and one Sushi (CCP/SCR) domain. (Ref.: Rauch, U., et al. (2001) Cell Mol. Life Sci. 58(12-13):1842-56; Zhou, XH et al. (2001). Mol. Cell Biol. 21(17): 5970-78).

Immunogen

GST-tagged recombinant fragement corresponding to 164 amino acids from the N-terminal region of rat Neurocan.

Application

Anti-Neurocan, N-terminal, Cat. No. ABT1384, is a rabbit polyclonal antibody that detects Neurocan core protein and has been tested for use in Immunofluorescence, Immunohistochemistry, and Radioimmunoassay.
Radioimmunoassay Analysis: A representative lot detected Neurocan in rat brain (Milev, P., et. al. (1998). Biochem Biophys Res Commun. 247(2):207-12).

Immunohistochemistry Analysis: A representative lot detected Neurocan in E13 and E14 mouse ventral diencephalon and retina in C57 mouse embryos (Leung, K.M., et. al. (2004). Brain Res Dev Brain Res. 152(1):1-10).


Quality

Evaluated by Immunofluorescence in E14 Embryonic rat brain (frozen section), rat E14 neocortex, and rat hippocampus cells.

Immunofluorescence Analysis: A 1:500 dilution of this antibody detected Neurocan in E14 Embryonic rat brain (frozen section), rat E14 neocortex, and rat hippocampus cells.

Target description

135.55 kDa calculated.

Physical form

Format: Unpurified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service