Skip to Content
Merck
All Photos(3)

Documents

ABT114

Sigma-Aldrich

Anti-LPA receptor 5 Antibody (GPR92)

from rabbit, purified by affinity chromatography

Synonym(s):

Lysophosphatidic acid receptor 5, LPA receptor 5, LPA-5, G-protein coupled receptor 92, G-protein coupled receptor 93

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LPAR5(57121)

General description

Lysophosphatidic acid receptor 5 (LPA receptor 5), also known as GPR92, is a G12/13 and Gq coupled receptor that mediates the signaling of LPA. Ligand binding to LPA receptor 5 may increase intracellular levels of cAMP and calcium, resulting in the activation of a wide range of effector molecules. Recent studies have demonstrated that LPA receptor 5 is abundantly expressed in human mast cells, and mediates the release of the macrophage inflammatory protein-1β (MIP-1β), indicating that LPA receptor 5 may play a role in innate and adaptive immunity.

Specificity

This antibody recognizes the extracellular domain of LPA receptor 5.

Immunogen

Epitope: Extracellular domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human LPA receptor 5.

Application

Detect the LPA receptor 5 protein using this Anti-LPA receptor 5 (GPR92) validated for use in WB.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
Western Blot Analysis: A representative lot detected LPA receptor 5 in non-transfected CHEM-1 cells and CHEM-1 cells transfected with LPAR5(GPR92) (Catalogue. No. HTS209C - Transfected Cell Line).

Immunocychemistry Analysis: A 1:500 dilution from a representative lot detected LPAR5 in TF1 cells, non-transfected CHEM-1 cells and CHEM-1 cells transfected with LPAR5(GPR92) (Catalogue. No. HTS209C - Transfected Cell Line).

NOTE: The protein used to make the transfected CHEM1 cell line is full length LPAR5.

Quality

Evaluated by Western Blot in K562 cell lysate.

Western Blot Analysis: 0.2 µg/mL of this antibody detected LPA receptor 5 on 10 µg of K562 cell lysate.

Target description

~37 kDa observed. The calculated molecular weight of this protein is 41 kDa, but can be observed at ~60 kDa due to high glycosylation.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisha Joshi et al.
International journal of molecular sciences, 22(16) (2021-08-28)
Increasing evidence suggests that systemic inflammation triggers a neuroinflammatory response that involves sustained microglia activation. This response has deleterious consequences on memory and learning capability in experimental animal models and in patients. However, the mechanisms connecting systemic inflammation and microglia
Camila O de Souza et al.
The Journal of clinical investigation, 132(21) (2022-09-07)
The molecular mechanisms underlying obesity-induced increases in β cell mass and the resulting β cell dysfunction need to be elucidated further. Our study revealed that GPR92, expressed in islet macrophages, is modulated by dietary interventions in metabolic tissues. Therefore, we
Maria Nunez-Salces et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 32(12), e13944-e13944 (2020-07-16)
The ability of the gut to detect nutrients is critical to the regulation of gut hormone secretion, food intake, and postprandial blood glucose control. Ingested nutrients are detected by specific gut chemosensors. However, knowledge of these chemosensors has primarily been
Feng Liu et al.
iScience, 26(8), 107465-107465 (2023-08-21)
Soluble epoxide hydrolase is a widely distributed bifunctional enzyme that contains N-terminal phosphatase (N-phos) and C-terminal epoxide hydrolase (C-EH) domains. C-EH hydrolyzes anti-inflammatory epoxy-fatty acids to corresponding diols and contributes to various inflammatory conditions. However, N-phos has been poorly examined. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service