Skip to Content
Merck
All Photos(2)

Documents

ABN481

Sigma-Aldrich

Anti-Synaptopodin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Synaptopodin, SYNPO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYNPO(11346)

General description

Synaptopodin (SYNPO) is an actin-associated protein that may play an important role in modulating actin-based shape and motility of dendritic spines and renal podocyte foot processes. It is expressed in the cerebral cortex. Mutations in SYNPO may be associated with nephrotic syndrome.

Immunogen

Epitope: Near C-terminus.
KLH-conjugated linear peptide corresponding to a sequence near the C-terminus of human SYNPO.

Application

Detect Synaptopodin using this rabbit polyclonal antibody, Anti-Synaptopodin Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: 2.5 µg/mL from a representative lot detected SYNPO in human brain tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected SYNPO in 15 µg of rat brain tissue lysate.

Target description

~95 kDa observed. Uniprot describes three isoforms at ~74 kDa, ~96 kDa, and ~97 kDa Uncharacterized bands may appear in some lysates.

Physical form

Affinity purified.
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hani Y Suleiman et al.
JCI insight, 2(16) (2017-08-18)
The architectural integrity of tissues requires complex interactions, both between cells and between cells and the extracellular matrix. Fundamental to cell and tissue homeostasis are the specific mechanical forces conveyed by the actomyosin cytoskeleton. Here we used super-resolution imaging methods
Yinghui Wang et al.
Renal failure, 46(2), 2381597-2381597 (2024-07-23)
Diabetic kidney disease (DKD) is one of the most common complications of diabetes. It is reported that mesenchymal stem cells (MSCs) derived exosomes (MSCs-Exo) may have great clinical application potential for the treatment of DKD, but the underlying mechanism has
Chunmei Xu et al.
Theranostics, 10(15), 6806-6824 (2020-06-20)
Diabetic nephropathy (DN) is mainly regarded as diabetic glomerulopathy, and its progression is tightly correlated with tubular epithelial lesions. However, the underlying molecular mechanisms linking tubular damage and glomerulopathy are poorly understood. Methods: We previously reported that the upregulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service