Skip to Content
Merck
All Photos(1)

Key Documents

ABN261

Sigma-Aldrich

Anti-POU6F1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

POU domain, class 6, transcription factor 1, Brain-specific homeobox/POU domain protein 5, Brain-5, Brn-5, mPOU homeobox protein, POU6F1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POU6F1(5463)

General description

POU domain, class 6, transcription factor 1 (POU6F1) is also called Brain-specific homeobox/POU domain protein 5 (Brn-5), and mPOU homeobox protein. POU6F1 is a nuclear transcription factor that binds preferentially to a variant of the octamer motif (5′-ATGATAAT-3′). POU6F1 is expressed exclusively in the developing embryonic brain, while in adults it is found in brain, heart, skeletal muscle and lung.

Specificity

This antibody recognizes the Gln/Pro rich domain of POU6F1.

Immunogen

Epitope: Gln/Pro rich domain
KLH-conjugated linear peptide corresponding to the Gln/Pro rich domain of human POU6F1.

Application

Anti-POU6F1 Antibody is a highly specific rabbit polyclonal antibody, that targets POU domain, class 6 & transcription factor 1 & has been tested in western blotting.

Quality

Evaluated by Western Blotting in human fetal brain tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected POU6F1 in 10 µg of human fetal brain tissue lysate.

Target description

~37 kDa observed. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service