Skip to Content
Merck
All Photos(2)

Documents

ABN2288

Sigma-Aldrich

Anti-Nesprin-2/SYNE2

Synonym(s):

KASH domain-containing protein 2, KASH2, Nuclear envelope spectrin repeat protein 2, Nucleus and actin connecting element protein, Protein NUANCE, Synaptic nuclear envelope protein 2, Syne-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

N/A, polyclonal

mol wt

calculated mol wt 782.75 kDa
observed mol wt ~135 kDa

species reactivity

mouse

species reactivity (predicted by homology)

canine

packaging

antibody small pack of 100 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

mouse ... Syne2(319565)

General description

Nesprin-2 (UniProt: Q6ZWQ0; also known as KASH domain-containing protein 2, KASH2, Nuclear envelope spectrin repeat protein 2, Nucleus and actin connecting element protein, Protein NUANCE, Synaptic nuclear envelope protein 2, Syne-2) is encoded by the Syne2 gene (Gene ID: 319565) in murine species. Nesprin-2 is a single-pass type IV membrane protein with a large cytoplasmic domain (aa 1-6823), a transmembrane domain (aa 6824-2844), and a perinuclear space domain (aa 6845-6874). It contains two calponin-homology domains (aa 31-136 and 183-288) and fifty-six spectrin repeats. Its actin-binding region is localized in amino acids 1-286. Nesprin-2 is a multi-isomeric modular protein that forms a linking network between organelles and the actin cytoskeleton to maintain the subcellular spatial organization. It is a core component of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex that is involved in the connection between the nuclear lamina and the cytoskeleton. The nucleocytoplasmic interactions established by the LINC complex play an important role in the transmission of mechanical forces across the nuclear envelope and in nuclear movement and positioning. Three different isoforms of Nesprin-2 have been described that are produced by alternative splicing. Different isoform patterns are found in the different compartments of the cell. Nesprin-2 levels are shown to be down-regulated in cancer cells. Nesprin-2 knockdown cells exhibit a defective nuclear translocation of c-Fos and Smad 2/3. Mutations in Syne2 gene have been linked to Emery-Dreifuss muscular dystrophy 5 (EDMD5), an autosomal dominant form of degenerative myopathy. (Ref.: Zhang, Q., et a; (2019). Curr. Biol. 29(17); 2826-2839; Kelkar, P., et al. (2015). Nucleus. 6(6); 479-489; Luxton, GWG., et al. (2010). Science. 329(5994); 956-959).

Specificity

This rabbit polyclonal antibody detects Nesprin-2. It targets an epitope within the N-terminal, actin-binding region.

Immunogen

GST-tagged recombinant fragment corresponding to 284 amino acids from the N-terminal, actin-binding region of mouse Nesprin-2.

Application

Quality Control Applications

Evaluated by Immunocytochemistry in NIH3T3 cells.

Immunocytochemistry Analysis: A 1:1,000 dilution of this antibody detected Nesprin-2/SYNE2 in NIH3T3 cells.

Tested applications

Western Blotting Analysis: A representative lot detected Nesprin-2/SYNE2 in Western Blotting application (Luxton, G.W., et. al. (2010). Science. 329(5994); 956-9).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Nesprin-2/SYNE2 in NIH-3T3 overexpressing Nesprin-2.

Immunocytochemistry Analysis: A representative lot detected Nesprin-2/SYNE2 in immunocytochemistry application (Zhang, Q., et. al. (2019). Curr Biol. 29(17); 2826-2839).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Physical form

Rabbit polyclonal antibody without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service