Skip to Content
Merck
All Photos(6)

Documents

ABN1381

Sigma-Aldrich

Anti-phospho-TrkB (Tyr816) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

BDNF/NT-3 growth factors receptor, Neurotrophic tyrosine kinase receptor type 2, TrkB tyrosine kinase, Trk-B, phospho-TrkB(Tyr816)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

species reactivity (predicted by homology)

gorilla (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology), human (based on 100% sequence homology), horse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr816 )

Gene Information

human ... NTRK2(4915)
rat ... Ntrk2(25054)

General description

BDNF/NT-3 growth factors receptor (EC 2.7.10.1; UniProt Q63604; also known as GP145-TrkB/GP95-TrkB, Trk-B, TrkB tyrosine kinase, Neurotrophic tyrosine receptor kinase type 2, Neural receptor protein-tyrosine kinase) is encoded by the Ntrk2 gene (also known as RATTRKB1, Trkb, TRKB1) in rat (Gene ID 25054). Neurotrophins mediate a wide range of neuronal functions, including survival, differentiation, growth, and death, via two classes of transmembrane receptors, the Trk receptor tyrosine kinase and the p75 neurotrophin receptor. While nerve growth factor (NGF), brain-derived neurotrophic factor (BDNF), neurotrophin-3 (NT-3), and NT-4 all bind the p75 receptor, TrkA is selective for NGF, TrkB is selective for BDNF and NT-4, and TrkC is selective for NT-3. TrkA and TrkB receptors can also be activated by ligands of the G-protein-coupled receptor (GPCRs), including the nucleoside adenosine or adenosine agonists, such as CGS 21680 and the pituitary adenylate cyclase-activating polypeptide (PACAP). In addition to transactivation via GPCRs and glucocorticoids are also reported to promote TrkA/B/C activation due to signaling cross-talk between Trk and GC receptor (GR).

Specificity

This polyclonal antibody selectively detects BDNF-induced pTyr816 phosphorylation of both glycosylated (145 kDa) and unprocessed (110 kDa) TrkB in primary embryonic rat E18 hippocampal neurons, but not NGF-induced pTyr490 or pTyr794 phosphorylation of TrkA in primary embryonic septal neurons (PMID 18347336).

Immunogen

Epitope: Near C-terminus
Linear peptide corresponding to rat TrkB sequence near the C-terminus with phosphorylated Tyr816.

Application

Detect phospho-TrkB (Tyr816) using this Anti-phospho-TrkB (Tyr816) Antibody validated for use in Western Blotting, Immunocytochemistry, Immunoprecipitation and Immunohistochemistry.
Immunocytochemistry Analysis: A representative lot detected BDNF- and adenosine-stimulated TrkB Tyr816 phosphorylation in cultured hippocampal neurons (Courtesy of Dr. Moses V. Chao, Langone Medical Center, New York University).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Tyr816 phosphorylated TrkB from BDNF-stimulated HEK293 cells expressing transfected TrkB (Courtesy of Dr. Moses V. Chao, Langone Medical Center, New York University).
Western Blotting Analysis: A representative lot detected dexamethasone-(Dex-) and BDNF-induced TrkB Tyr816 phosphorylation in cortical slices from postnatal P9–P10 rats, as well as in dorsal striatum from P18-20 rats received intraperitoneal Dex injection (Jeanneteau, F., et al. (2008). Proc Natl Acad Sci USA. 105(12):4862-4867).
Western Blotting Analysis: A representative lot detected TrkB Tyr816 phosphorylation in mice brain synaptosome preparations (Parkhurst, C.N., et al. (2013). Cell. 155(7):1596-1609).
Western Blotting Analysis: A representative lot detected BDNF-stimulated TrkB Tyr816 phosphorylation in cultured mouse hippocampal neurons and HEK293 cells expressing murine TrkB (Bath, K.G., et al. (2008). J. Neurosci. 28(10):2383-2393).
Immunohistochemistry Analysis: A representative lot detected enhanced TrkB Tyr816 phosphorylation in the dentate gyrus tissue sections from postnatal P18 rats after receiving metyrapone and/or dexamethasone treatment (Jeanneteau, F., et al. (2008). Proc Natl Acad Sci USA. 105(12):4862-4867).
Immunohistochemistry Analysis: A representative lot detected TrkB Tyr816 phosphorylation in coronal slices of the rostral migratory stream (RMS) from adult mice (Bath, K.G., et al. (2008). J. Neurosci. 28(10):2383-2393).
Immunocytochemistry Analysis: A representative lot detected corticosterone-stimulated TrkB Tyr816 phosphorylation in 5-hour starved rat hippocampal neuron culture (Jeanneteau, F., et al. (2008). Proc Natl Acad Sci USA. 105(12):4862-4867).
Immunocytochemistry Analysis: A representative lot detected BDNF-stimulated TrkB Tyr816 phosphorylation in cultured DRG neurons from E16.5 rat embryos (Arevalo, J.C., et al. (2006). Neuron. 50(4):549-559).
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in BDNF treated murine TrkB-transfected HEK293 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected phospho-TrkB (Tyr816) in 10 µg of BDNF treated murine TrkB-transfected HEK293 cell lysate.

Target description

~140 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanjun Liu et al.
Redox biology, 43, 102002-102002 (2021-05-14)
Emerging evidence suggests that the reduction of ethanolamine plasmalogen (PlsEtn) is associated with in Alzheimer's disease and metabolic diseases. However, the mechanistic bases for PlsEtn on the these diseases are not well understood. Plasmalogens are primarily synthesized in the liver
Dong Wook Lim et al.
Antioxidants (Basel, Switzerland), 10(8) (2021-08-28)
Menopause is a risk factor for depression. Although 1,3-dicaffeoylquinic acid (1,3-diCQA), a phenolic compound in Arctium lappa (A. lappa) root, has various health benefits, its effects on menopausal depression remain to be determined. Therefore, this study investigates the antidepressant-like effects
Weixing Zhao et al.
Journal of neuroinflammation, 16(1), 230-230 (2019-11-22)
It is widely accepted that mitochondria have a direct impact on neuronal function and survival. Oxidative stress caused by mitochondrial abnormalities play an important role in the pathophysiology of lipopolysaccharide (LPS)-induced memory impairment. Elamipretide (SS-31) is a novel mitochondrion-targeted antioxidant.
Guangdong Chen et al.
Biomedical reports, 6(3), 285-290 (2017-04-30)
The aim of the present study was to investigate the effects of a commonly-used atypical antipsychotic, risperidone, on alterations in spatial learning and in the hippocampal brain-derived neurotrophic factor (BDNF)-tyrosine receptor kinase B (TrkB) signalling system caused by acute dizocilpine
Pavel A Kipnis et al.
JCI insight, 5(12) (2020-05-20)
Refractory neonatal seizures do not respond to first-line antiseizure medications like phenobarbital (PB), a positive allosteric modulator for GABAA receptors. GABAA receptor-mediated inhibition is dependent upon electroneutral cation-chloride transporter KCC2, which mediates neuronal chloride extrusion and its age-dependent increase and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service