Skip to Content
Merck
All Photos(2)

Key Documents

ABF188

Sigma-Aldrich

Anti-IRF-1

from rabbit, purified by affinity chromatography

Synonym(s):

Interferon regulatory factor 1, IRF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... IRF1(3659)

General description

Interferon regulatory factor 1 (UniProt: P10914; also known as IRF-1) is encoded by the IRF1 gene (Gene ID: 3659) in human. IRF-1 is a transcriptional regulator that is expressed constitutively in all cells types except early embryonal cells. It can exist as a monomer and as a homodimer. It displays remarkable functional diversity in the regulation of cellular responses, including the regulation of interferon (IFN) and IFN-inducible genes, host response to viral and bacterial infections. It also regulates many genes expressed during hematopoiesis, inflammation, immune responses, cell proliferation and differentiation, and growth arrest and apoptotic cell death following DNA damage. IRF-1 is reported to stimulate p53/TP53-dependent transcription through enhanced recruitment of EP300 leading to increased acetylation of p53/TP53. Its DNA-binging domains are located in the first 115 amino acids of N-terminal region and contains a series of five well-conserved tryptophan-rich repeats. IRF-1 can be phosphorylated by casein kinase 2, which positively regulates its activity. Mutations in IRF1 gene are reported to cause gastric cancer. (Ref.: Dou, L., et al. (2014). Hum. Immunol. 75(11); 1110-1114).

Specificity

This rabbit polyclonal antibody detects Interferon Regulaotry Factor 1 in human cells. It targets an epitope within 14 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal half of human Interferon Regulatory Factor 1.

Application

Anti-IRF-1, Cat. No. ABF188, is a rabbit polyclonal lantibody that detects Interferon regulatory factor 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Inflammation & Immunology
Western Blotting Analysis: A 1:500 dilution from a representative lot detected IRF-1 in Jurkat cell lysate.

Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected IRF-1 in human stomach cancer and human small intestine tissue sections.

Quality

Evaluated by Western Blotting in PC12 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected IRF-1 in PC12 cell lysate.

Target description

~48 kDa observed; 36.50 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service