ABF1820
Anti-Influenza A nucleoprotein
serum, from rabbit
Synonym(s):
Nucleoprotein, Nucleocapsid protein, Protein N
About This Item
Recommended Products
biological source
rabbit
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
virus, influenza A virus
packaging
antibody small pack of 25 μL
technique(s)
immunofluorescence: suitable
western blot: suitable
isotype
IgG
GenBank accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
SARS coronavirus ... N(1489678)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A 1:3,000 dilution from a representative lot detected Influenza A nucleoprotein in A549 cells were infected with A/PR/8/34 strain (Courtesy of Dr. Amelia Nieto at CNB).
Western Blotting Analysis: A representative lot detected Influenza A nucleoprotein in Western Blotting applications (Jorba, N., et. al. (2009). PLoS Pathog. 5(5):e1000462; Lukarsk, M., et. al. (2017). Nature. 541(7635):117-121; Tripathi, S., et. al. (2015). Cell Host Microbe. 18(6):723-35).
Immunofluorescence Analysis: A representative lot detected Influenza A nucleoprotein in Immunofluorescence applications (Marcos-Villar, L., et. al. (2018). Sci Rep. 8(1):1230; Rodriguez-Fandsen, A., et. al. (2016). Sci Rep. 6:20744).
Inflammation & Immunology
Quality
Western Blotting Analysis: A 1:5,000 dilution of this antibody detected Influenza A nucleoprotein in A549 cells infected with A/Victoria/3/75 strain of influenza virus.
Target description
Physical form
Storage and Stability
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service