Skip to Content
Merck
All Photos(2)

Key Documents

ABE604

Sigma-Aldrich

Anti-Bonzo Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, CDw186, G-protein coupled receptor STRL33, G-protein coupled receptor bonzo, CD186, BONZO, STRL33, TYMSTR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CXCR6(10663)

General description

G-protein coupled receptor bonzo (BONZO), also known as C-X-C chemokine receptor type 6 (CXCR6,CXC-R6,CXCR-6), CDw186 (CD186), G-protein coupled receptor STRL33 (STRL33), and TYMSTR, is a receptor for the C-X-C chemokine CXCL16 and is shown to be used as a coreceptor by SIVs and by strains of HIV-2 and m-tropic HIV-1. Certain G protein-coupled receptors including CCR5, CXCR4, CCR3, and CCR2b have been identified as HIV coreceptors. BONZO is expressed in lymphoid tissues and activated T cells.

Specificity

This antibody recognizes the N-terminus of Bonzo.

Immunogen

KLH-conjugated linear peptide corresponding to the N-terminus of human Bonzo.

Application

Detect CXCR6 using this rabbit polyclonal antibody, Anti-Bonzo Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: 20 µg/mL from a representative lot detected Bonzo in human spleen tissue.

Quality

Evaluated by Western Blotting/Peptide Inhibition Assay in human spleen tissue lysate.

Western Blotting/Peptide Inhibition Assay Analysis: 1 µg/mL of this antibody detected Bonzo in 15 µg of human spleen tissue lysate.

Target description

~45 kDa observed. Uncharacterized bands may appear in some lysate(s).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service