Skip to Content
Merck

Skip To

ABE577

Anti-Myb Binding Protein 1A Antibody

serum, from rabbit

Synonym(s):

Myb-binding protein 1A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, ICC, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ChIP: suitable, immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYBBP1A(10514)

General description

Myb-binding protein 1A (MYBBP1A) interacts with sequence specific DNA-binding proteins in order to activate or repress transcription. Transcriptional repression via Myb-binding protein 1A is thought to be mediated by histone deacetylase activity (HDAC activity).
~160 kDa observed. Uncharacterized band(s) may be observed in some cell lysates. The full length of this protein is observed at ~160 kDa; however, it is proteolytically cleaved into ~140 kDa and ~67 kDa fragments (Hochstatter, J., et al. (2012). J Biol Chem. 287(29):24365-24377.; Mori, S., et al. (2012). PLoS One. 7(10):e39723.).

Immunogen

GST-tagged recombinant protein corresponding to human Myb Binding Protein 1A.

Application

Anti-Myb Binding Protein 1A Antibody is a highly specific rabbit polyclonal antibody, that targets Myb-binding protein 1A & has been tested in western blotting & ICC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Myb Binding Protein 1A in diploid human lung embryonic fibroblasts (Prof. Atilla Nemeth, University of Regensburg, Germany.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected Myb Binding Protein 1A in HeLa cells (Hochstatter, J., et al. (2012). J Biol Chem. 287(29):24365-24377.).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot from an independent laboratory immunoprecipitated Myb Binding Protein 1A in ChIP (Hochstatter, J., et al. (2012). J Biol Chem. 287(29):24365-24377.).

Biochem/physiol Actions

This antibody detects the full length and cleaved forms of Myb Binding Protein 1A.

Analysis Note

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analyis: A 1:1,000 dilution from a representative lot detected Myb Binding Protein 1A in 10 µg of A431 cell lysate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABE886ABE1330ABE262
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

antibody form

serum

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

serum

UniProt accession no.

Q9BQG0

UniProt accession no.

P10244

UniProt accession no.

Q96AE4

UniProt accession no.

P56524

species reactivity

human

species reactivity

mouse, human

species reactivity

human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ABE57704053252913952

Questions

Reviews

No rating value

Active Filters