Skip to Content
Merck
All Photos(2)

Key Documents

ABE552

Sigma-Aldrich

Anti-Alpha-ketoglutarate-dependent dioxygenase FTO Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Alpha-ketoglutarate-dependent dioxygenase FTO, Fat mass and obesity-associated protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), sheep (based on 100% sequence homology), ox (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FTO(79068)

General description

Alpha-ketoglutarate-dependent dioxygenase FTO (Fat mass and obesity-associated protein) is a ubiquitous nuclear protein which belongs to the FTO family. It is a dioxygenase enzyme that dealkylates nucleic acids by oxidative demethylation in the presence of oxygen, alpha-ketoglutarate and iron (Fe2+). FTO is highly active on single-stranded RNA molecules with 3-methyluracil residues and on single-stranded DNA containing 3-methylthymine residues, and has relatively low activity on single-stranded DNA containing either 3-methylcytosine or 3-methyladenine. There has been no observed activity on double-stranded DNA. FTO is highly abundant in the hypothalamus, pituitary, and adrenal glands. It plays an important role in energy homeostasis and the regulation of fat accumulation in the body, and may be involved in obesity. Mutated FTO proteins are linked to a syndrome characterized by retarded growth, delayed development, coarse facies and early death.

Immunogen

KLH-conjugated linear peptide corresponding to human Alpha-ketoglutarate-dependent dioxygenase FTO.

Application

Immunoprecipitation Analysis: 10 μg from a representative lot immunoprecipitated Alpha-ketoglutarate-dependent dioxygenase FTO from 500 μg of U2OS nuclear extract.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
This Anti-Alpha-ketoglutarate-dependent dioxygenase FTO Antibody is validated for use in western blotting & IP for the detection of Alpha-ketoglutarate-dependent dioxygenase FTO.

Quality

Evaluated by Western Blotting in U2OS nuclear extract.

Western Blotting Analysis: 1 μg/mL of this antibody detected Alpha-ketoglutarate-dependent dioxygenase FTO in 10 μg of U2OS nuclear extract.

Target description

~58 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Liang Ke et al.
Bioengineered, 13(3), 5443-5452 (2022-02-19)
Reperfusion therapy after acute myocardial infarction can induce myocardial ischemia-reperfusion injury (IRI). Novel evidence has illustrated that N6-methyladenosine (m6A) modification modulates the myocardial IRI progression. Here, our study focuses on the role of m6A methyltransferase fat mass and obesity-associated protein
Bo Qin et al.
Oncology reports, 45(3), 1059-1071 (2021-03-03)
Long non‑coding RNAs (lncRNAs) are involved in the regulation of esophageal squamous cell carcinoma (ESCC) progression. However, the function and mechanism of lncRNA cancer susceptibility candidate 15 (CASC15) are poorly defined. In the present study, tumor and normal adjacent tissues were collected from
John P Zepecki et al.
PLoS genetics, 17(3), e1009086-e1009086 (2021-03-09)
Within the glioblastoma cellular niche, glioma stem cells (GSCs) can give rise to differentiated glioma cells (DGCs) and, when necessary, DGCs can reciprocally give rise to GSCs to maintain the cellular equilibrium necessary for optimal tumor growth. Here, using ribosome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service