Skip to Content
Merck
All Photos(1)

Key Documents

ABE479

Sigma-Aldrich

Anti-hMOF Antibody

from rabbit

Synonym(s):

Histone acetyltransferase KAT8, Lysine acetyltransferase 8, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST-1, hMOF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KAT8(84148)

General description

hMOF, also known as Histone acetyltransferase KAT8, or Lysine acetyltransferase, MOZ, YBF2/SAS3, SAS2, TIP60 protein 1, and MYST-1, and encoded by the gene KAT8/MOF/MYST1, is a histone acetyltransferase which is involved in transcriptional activation and also is a component of the MSL complex involved in acetylation of nucleosomal histone H4 producing specifically H4K16ac. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. hMOF also a component of many other MSL related complexes including the methyltransferase complexes and interacts with numerous other related proteins including BAP18, CHD8, E2F6, HSP70, INO80C, KANSL1, LAS1L, MAX, MCRS1, MGA, MOF/KAT8, PELP1, PHF20, PRP31, RING2, RUVB1/TIP49A, RUVB2/TIP49B, SENP3, TAF1, TAF4, TAF6, TAF7, TAF9 and TEX10. hMOF is localized to the nucleus and nuclear chromosomes.

Immunogen

Linear peptide corresponding to hMOF.

Application

Anti-hMOF antibody detects level of hMOF & has been published & validated for use in hMOF.
Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated hMOF from HeLa cell lysate (Wu, L., et al. (2011). Mol Cell. 43(1):132-144.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated hMOF from HeLa cell lysate (Smith, E. R., et al. (2005). Mol Cell Biol. 25(21):9175-9188.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HUVEC cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected hMOF in 10 µg of HUVEC cell lysate.

Target description

~60 kDa observed. Uncharacterized band(s) may be observed in some cell lysate.

Physical form

Format: Purified
Protein G Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service