Skip to Content
Merck
All Photos(5)

Key Documents

ABE290

Sigma-Aldrich

Anti-TET3

from rabbit, purified by affinity chromatography

Synonym(s):

Methylcytosine dioxygenase TET3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), rat (based on 100% sequence homology), nonhuman primates (based on 100% sequence homology)

technique(s)

ChIP: suitable (ChIP-seq)
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TET3(200424)

General description

MABE1144/1132/1133 Methylcytosine dioxygenase TET3 (EC 1.14.11.n2; UniProt O43151) is encoded by the TET3 (also known as KIAA0401) gene (Gene ID 200424) in human. TET (ten-eleven translocation) proteins (TET1, TET2, and TET3) oxidize 5-methylcytosine (5mC) to 5-hydroxymethylcytosine (5hmC), 5-formylcytosine (5fC), and 5-carboxylcytosine (5caC), providing a means for active epigenetic DNA demethylation in mammals. TET proteins also modify genomic thymine residues to 5-hydroxyuracil. TET proteins depend on Fe(II) and 2-oxoglutarate as cofactors for activity, and 2-oxoglutarate is known to inhibit their activity, while ascorbate is shown to stimulate TET-mediated cytosine oxidation. In addition, post-translational modifications (PTMs), such as phosphorylation and O-GlcNAcylation, also play a role in regulating the activity of TET proteins. TET1 and TET2 are found highly expressed in mouse embryonic stem cells (mESCs) and TET3 is known to be upregulated in oocytes and oxidize the silenced paternal pronuclear DNA. High levels of TET proteins and genomic 5-hydroxymethylcytosine in neuronal tissues are also reported. Three TET3 spliced isoforms/variants have been reported, the long or full-length isoform Tet3FL and the short isoform Tet3s are upregulated during embryonic stem cell (ESC) differentiation toward the neuronal lineage, while Tet3o is oocyte-specific and not detected in ESCs or in any other cell types or adult mouse tissues tested, indicating a role of Tet3o in paternal genome oxidation in mouse zygotes. Tet3FL and Tet3o contain the entire Tet3s sequence with additional (but different) N-terminal sequence. Only Tet3FL contains an N-terminal CXXC domain that binds to unmethylated CpGs and exhibits highest affinity toward 5caC. Tet3FL is substantially less active than Tet3o and Tet3s, suggesting that the CXXC domain restricts Tet3FL 5mC oxidation capacity.

Specificity

Immunogen sequence is present in TET3 isoforms Tet3FL, Tet3s, and Tet3o spliced isoforms reported by Jin, S.G., et al. (2016). Cell Rep. 14(3):493-505), as well as all additional human and mouse TET3 spliced isoforms reported by UniProt (O43151, Q8BG87). Note that UniProt isoform 1 (O43151-1/human and Q8BG87-1/mouse) corresponds to isoform Tet3s, not Tet3FL (NM_001287491.1/human and JX946278.1/mouse).

Immunogen

KLH-conjugated linear peptide corrsponding to an internal sequence within the N-terminal half of human TET3.

Application

Anti-TET3, Cat. No. ABE290, is a highly specific rabbit polyclonal antibody that targets methylcytosine dioxygenase TET3 and has been tested in Chromatin Immunoprecipitation (ChIP), ChIP-seq, Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated the exogenously expressed mouse TET3-GFP fusion protein from 500 µg lysate of transfected mouse embryonic stem cells (mESCs).

Chromatin Immunoprecipitation-sequencing (ChIP-seq) Analysis: A representative lot detected TET3 chromosome enrichment sites by ChIP-seq analysis (10 µg per ChIP) using chromatin preparation from mouse embryonic stem cells (mESCs) transfected to overexpress mouse TET3 (Courtesy of Reik lab, Epigenetics ISP, Babraham Institute, UK).

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot immunostained 4% paraformaldehyde-fixed, 0.5% Triton X-100-permeabilized mouse embryonic stem cells (mESCs) transfected to overexpress mouse TET3 oocyte isoform Tet3o (Courtesy of Reik lab, Epigenetics ISP, Babraham Institute, UK).

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected the immunoreactivity of TET3 oocyte isoform Tet3o in 4% paraformaldehyde-fixed, 0.5% Triton X-100-permeabilized mouse zygotes (Courtesy of Reik lab, Epigenetics ISP, Babraham Institute, UK).

Immunoprecipitation Analysis: 10 µg from a representative lot co-immunoprecipitated endogenous OGT with the exogenously overexpressed mouse TET3 from transfected mouse embryonic stem cells (mESCs) by RIME (Rapid Immunoprecipitation Mass spectrometry of Endogenous proteins) (Courtesy of Reik lab, Epigenetics ISP, Babraham Institute, UK).

Quality

Evaluated by Western Blotting of mouse TET3 GFP fusion.

Western Blotting Analysis: 2 µg/mL of this antibody detected the exogenously expressed mouse TET3 GFP fusion protein in 10 µg of lysate from transfected mouse embryonic stem cells (mESCs).

Target description

193.7/179.4 kDa (human isoform Tet3FL/Tet3s), 194.8/180.4/185.1 kDa (mouse isoform Tet3FL/Tet3s/Tet3o), 166.7/77.47 kDa (UniProt human isoform 2/3), 76.26 kDa (UniProt mouse isoform 2) calculated. Note that UniProt isoform 1 (O43151-1/human and Q8BG87-1/mouse) corresponds to isoform Tet3s, not Tet3FL (NM_001287491.1/human and JX946278.1/mouse).

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt..

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Li et al.
Breast cancer research : BCR, 26(1), 44-44 (2024-03-12)
Ubiquitin carboxyl-terminal hydrolase L1 (UCHL1) is a deubiquitinating enzyme that regulates ERα expression in triple-negative cancer (TNBC). This study aimed to explore the deubiquitination substrates of UCHL1 related to endocrine therapeutic responses and the mechanisms of UCHL1 dysregulation in TNBC.
Harmony C Ketchum et al.
Communications biology, 7(1), 415-415 (2024-04-06)
The ten-eleven-translocation family of proteins (TET1/2/3) are epigenetic regulators of gene expression. They regulate genes by promoting DNA demethylation (i.e., catalytic activity) and by partnering with regulatory proteins (i.e., non-catalytic functions). Unlike Tet1 and Tet2, Tet3 is not expressed in
Aixia Liu et al.
iScience, 24(9), 103065-103065 (2021-09-28)
A conceptual framework for understanding abnormal endometrial decidualization, with considerable significance for the diagnosis and treatment of abnormal decidualization-related changes in non-receptive endometrium in implantation failure during early pregnancy is very important. Here, we found the expression levels of miR-29a
A small-molecule degrader of TET3 as treatment for anorexia nervosa in an animal model.
Lv, et al.
Proceedings of the National Academy of Sciences of the USA, 120, e2300015120-e2300015120 (2023)
Tiefeng Cao et al.
Oncogene, 38(27), 5356-5366 (2019-05-16)
Uterine leiomyomas or fibroids (UFs) are benign tumors characterized by hyperplastic smooth muscle cells and excessive deposition of extracellular matrix (ECM). Afflicting ~80% of women, and symptomatic in 25%, UFs bring tremendous suffering and are an economic burden worldwide; they

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service