Skip to Content
Merck
All Photos(2)

Documents

ABE281

Sigma-Aldrich

Anti-EZH1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone-lysine N-methyltransferase EZH1, ENX-2, Enhancer of zeste homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), bovine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EZH1(2145)

General description

Polycomb protein group complexes are responsible for the regulation of hundreds of genes in mammals and insects. These proteins are responsible for the assembly and packaging of chromatin and for the induction of methylation on Histone H3. EZH1 is the catalytic subunit of the PRC2/EED-EZH1 complex. This complex methylates Lys-27 of histone H3, which leads to transcriptional repression of the affected target gene. EZH1 is required for embryonic stem cell derivation and self-renewal, suggesting that it is involved in maintaining the identity of embryonic stem cells.

Specificity

Other homologies: Porcine (94% sequence homology).
This antibody potentially recognizes all isoforms of EZH1.

Immunogen

KLH-conjugated linear peptide corresponding to human EZH1.

Application

Detect the EZH1 protein using this Anti-EZH1 validated for use in IHC & WB.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected EZH1 in human lung and kidney tissues.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins

Quality

Evaluated by Western Blot in mouse embryonic stem cell lysate.

Western Blot Analysis: 0.2 µg/mL of this antibody detected EZH1 in 10 µg of mouse embryonic stem cell lysate.

Target description

~85 kDa observed. Uniprot states 5 isoforms for EZH1. This antibody potentially recognizes all isoforms. Isoform A is 85,271 Daltons. Isoform B is 86,025 Daltons. Isoform C is 80,699 Daltons. Isoform D is 76,952 Daltons. Isoform E is 68,961 Daltons. (Margueron, 2008).

Linkage

Replaces: 04-1047

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse embryonic stem cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian C B Bremer et al.
Digestion, 102(2), 227-235 (2019-11-07)
Colorectal cancer (CRC) is the leading gastrointestinal malignancy. The development from premalignant intraepithelial lesions leading to invasive cancer is paradigmatic for the stepwise carcinogenesis of epithelial cancers, but the knowledge of the underlying mechanism of carcinogenesis and progression of CRC
Jing Liang et al.
British journal of pharmacology, 176(17), 3206-3219 (2019-06-05)
The increased proliferation and migration of vascular smooth muscle cells (VSMCs) after arterial injury contributes greatly to the pathogenesis of neointimal hyperplasia. As a major component of epigenetics, histone methylation plays an important role in several cardiovascular diseases. However, its
Sebastian C B Bremer et al.
Cancers, 14(12) (2022-06-25)
Tumor grading is a robust prognostic predictor in patients with neuroendocrine neoplasms (NEN) and guides therapy, especially in tumors with high proliferation. NEN can be separated into well-differentiated and poorly differentiated types. The more aggressive NEN have been further separated
Jiajia Wang et al.
Science advances, 6(33), eaaz6477-eaaz6477 (2020-08-28)
Mutations in the polycomb repressive complex 2 (PRC2) can cause Weaver-like syndrome, wherein a patient cohort exhibits abnormal white matter; however, PRC2 functions in CNS myelination and regeneration remain elusive. We show here that H3K27me3, the PRC2 catalytic product, increases
Xue-Wei Wang et al.
The Journal of clinical investigation, 134(3) (2023-11-28)
Current treatments for neurodegenerative diseases and neural injuries face major challenges, primarily due to the diminished regenerative capacity of neurons in the mammalian CNS as they mature. Here, we investigated the role of Ezh2, a histone methyltransferase, in regulating mammalian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service