Skip to Content
Merck
All Photos(3)

Key Documents

ABE269

Sigma-Aldrich

Anti-acetyl-Histone H2B (Lys24) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H2B type 3-B, H2B type 12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

technique(s)

dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (pLys24)

Gene Information

human ... H2BC1(255626)
mouse ... H2Bc1(319177)
rat ... H2Bc1(24829)

General description

Changes in chromatin structure play a large role in the regulation of transcription in eukaryotes (1). The nucleosome is the primary building block of chromatin, and is made up of four core histone proteins (H2A, H2B, H3 and H4) (2). Acetylation of core histones regulates gene expression (2). Histone H2B is primarily acetylated at lysines 5, 12, 15, and 20 (3). Ubiquitylation of Histone H2B at lysine 123 by Rad6 has been implicated in transcriptional activation and lysine methylation on histone H3 (4). Histone H2B phosphorylation at Ser10 (for yeast) and Ser14 (for mammalian cells) has been linked to regulation of apoptosis and meiosis (5).

Specificity

This antibody recognizes Histone H2B acetylated at Lys24.

Immunogen

Epitope: Acetylated Lys24
KLH-conjugated linear peptide corresponding to human Histone H2B acetylated at Lys24.

Application

Immunoprecipitation Analysis: 10 μg from a representative lot immunoprecipitated acetyl-Histone H2B (Lys24) from HeLa acid extract.

Dot Blot Specificity Analysis: Unmodified and various modified Histone peptides (see Table) were probed with Anti-acetyl-Histone H2B (Lys24) (1:500 dilution).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-acetyl-Histone H2B (Lys24) Antibody (Rabbit Polyclonal Antibody) validated in WB, IP, DB to detect acetyl-Histone H2B (Lys24) also known as H2BK24Ac, Histone H2B (acetyl K24).

Quality

Evaluated by Western Blot in recombinant Histone H2B protein, and untreated and sodium butyrate treated HeLa acid extracts.

Western Blot Analysis: A 1:500 dilution of this antibody detected Histone H2B on 10 µg of recombinant Histone H2B protein, and untreated and sodium butyrate treated HeLa acid extracts.

Target description

~16 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Recombinant Histone H2B protein, and untreated and sodium butyrate treated HeLa acid extracts

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service