Skip to Content
Merck
All Photos(3)

Key Documents

ABE184

Sigma-Aldrich

Anti-Fox2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

RNA binding protein fox-1 homolog 2, Fox-1 homolog B, Fox-1 homolog Fxh, Hexaribonucleotide-binding protein 2, RNA-binding motif protein 9, RNA-binding protein 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RBFOX2(23543)

General description

Fox2, also referred to as RNA binding protein fox-1 homolog 2, is an RNA binding protein involved in the regulation of tissue-specific alternative splicing. This protein interacts with the ER-alpha N-terminal activation domain, and contains a highly conserved RRM (RNA recognition motif), which associates with 5′-UGCAUGU-3′ elements. This protein has been demonstrated to regulate alternative splicing of tissue specific exons and differentially spliced exons during erythropoiesis.

Specificity

Other homologies: Bovine (98% sequence homology), Canine (98% sequence homology), Equine (98% sequence homology).

Immunogen

MBP-tagged recombinant protein corresponding to mouse Fox2.

Application

Immunohistochemistry Analysis: A 1:300 dilution from a representative lot detected Fox2 in human cerebellum tissue.

Immunocytochemistry Analysis: 1 µg/mL from a representative lot detected Fox2 in A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-Fox2 Antibody (Rabbit Polyclonal Antibody) validated in IHC(P), ICC, WB to detect Fox2 also known as RNA binding protein fox-1 homolog 2, Fox-1 homolog B.

Quality

Evaluated by Western Blot in C2C12 cell lysate.

Western Blot Analysis: 50 ng/mL of this antibody detected Fox2 in 10 µg of C2C12 cell lysate.

Target description

~48 kDa observed. Alternative splicing can result in up to 9 isoforms at various molecular weights. Uncharacterized bands between ~60-80 kDa have been observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
C2C12 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service