Skip to Content
Merck
All Photos(3)

Documents

ABE1425

Sigma-Aldrich

Anti-COUP-TF1 Antibody

serum, from rabbit

Synonym(s):

COUP transcription factor 1, COUP-TF1, COUP transcription factor I, COUP-TF I, Nuclear receptor subfamily 2 group F member 1, V-erbA-related protein 3, EAR-3, COUP-TF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

ChIP: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Nr2F1(13865)
rat ... Nr2F1(81808)

General description

COUP transcription factor 1 (UniProt Q60632; also known as Avian erythroblastic leukemia viral oncogene homolog-like 3, COUP-TF1, COUP-TF I, COUP transcription factor I, EAR-3, Nuclear receptor subfamily 2 group F member 1, V-erbA-related protein 3) is encoded by the Nr2f1 (also known as Erbal3, Tfcoup1) gene (Gene ID 13865) in murine species. COUP transcription factors1&2 (COUP-TFI & COUP-TFII) are two related nuclear orphan receptors implicated in their roles in guiding migrating neurons towards their target regions during brain development. COUP-TFI and COUP-TFII are expressed in distinct and non-overlapping migrating cells in specific pathways in the basal telencephalon of mouse embryos at a stage when interneuron migration is robust. While COUP-TFI appears to control dorsal and ventral migration in the basal telencephalon, COUP-TFII is more specific in modulating tangential migration into the intermediate zone of the cortex. Coup-TFII is also identified as an activator of Prox1 expression in venous lymphatic endothelial cell (LEC) progenitors during early stages of LEC specification and differentiation.

Specificity

Specificity of the antiserum was confirmed by immunogen blocking in immunohistochemistry on frozen sections (Tripodi, M., et al. (2004). Development. 131(24):6119-6129).

Immunogen

Epitope: N-terminal half
GST-tagged recombinant protein corresponding to the N-terminal half of mouse COUP-TF1.

Application

Anti-COUP-TF1 Antibody is an antibody against COUP-TF1 for use in Western Blotting, Immunohistochemistry, Immunofluorescence, Chromatin Immunoprecipitation (ChIP).
Immunohistochemistry Analysis: An 1:250 dilution from a representative lot detected COUP-TFI in 20 µg of rat cerebral cortex, human cerebellum, and human small intestine tissue.
Immunofluorescence Analysis: An 1:500-1000 dilution of a representative lot detected COUP-TF1 immunoreactivity in the developing brain using paraformaldehyde-fixed cryosections from wild-type mouse E13.5 embryos, but not in embryos with brain region-specific COUP-TF1-knockout, by fluorescent immunohistochemistry (Courtesy of Dr. Michele Studer, Institut de Biologie Valrose, France).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected COUP-TF1 occupancy at the conserved binding sites along the Rnd2 locus using chromatin preparations from the neocortices of E14.5 mouse embryos (Alfano, C., et al. (2011). Development. 138(21):4685-4697).
Western Blotting Analysis: A representative lot detected a higher COUP-TF1 level in retinal and retinal pigment epithelium (RPE) extracts from 24-week-old mice subjected to prorolonged light exposure (24 h/day) than from mice with normal light exposure (12 h/day). Light stress-induced COUP-TF1 upregulation was also observed in RPE, but not retinal extracts from Ranbp2+/− mice (Cho, K.I., et al. (2010). Dis. Model Mech. 3(9-10):595-604).
Western Blotting Analysis: A representative lot detected a 64 kDa form of COUP-TF1 co-immunoprecipitated with RNABP2 from murine retinal extracts (Cho, K.I., et al. (2010). Dis. Model Mech. 3(9-10):595-604).
Immunofluorescence Analysis: A representative lot detected cortex-specific COUP-TFI-knockout in the developing brain of E11.5 COUP-TFIfl/flEmx1Cre transgenic mouse embryos by fluorescent immunohistochemistry staining of paraformaldehyde-fixed cryosections (Armentano, M., et al. (2007). Nat. Neurosci. 10(10):1277-1286).
Immunofluorescence Analysis: A representative lot detected A representative lot detected COUP-TFI immunoreactivity in paraformaldehyde-fixed frozen coronal sections from wild-type E15.5 mouse embryos, but not from COUP-TFI-knockout embryos (Armentano, M., et al. (2006). Development. 133(21):4151-4162).
Immunofluorescence Analysis: A representative lot detected COUP-TF1 immunoreactivity associated with calbindin-positive GABAergic interneurons, but not reelin-positive Cajal-Retzius cells in the developing cortex by dual fluorescent immunohistochemistry using cryosections from E13.5 and E15.5 mouse embryos (Tripodi, M., et al. (2004). Development. 131(24):6119-6129).
Immunohistochemistry Analysis: A representative lot detected a different COUP-TF1 expression pattern than that of COUP-TF2 in the developing telencephalon of forebrain using cryosections from E13.5 mouse embryos (Tripodi, M., et al. (2004). Development. 131(24):6119-6129).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected COUP-TFI in 20 µg of mouse brain tissue lysate.

Target description

~50-55kDa kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal antibody in serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Rosebrock et al.
Nature cell biology, 24(6), 981-995 (2022-06-14)
Cerebral organoids exhibit broad regional heterogeneity accompanied by limited cortical cellular diversity despite the tremendous upsurge in derivation methods, suggesting inadequate patterning of early neural stem cells (NSCs). Here we show that a short and early Dual SMAD and WNT
Qi Xing et al.
Cell regeneration (London, England), 8(1), 1-8 (2019-06-18)
Retrograde monosynaptic tracing with EnvA-pseudotyped rabies virus has been employed to identify the afferent and efferent connectivity of transplanted human embryonic stem (hES) cell-derived neurons in animal models. Due to the protracted development of transplanted human neurons in host animals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service