Skip to Content
Merck

Skip To

ABE1024

Anti-Oligodendrocyte transcription factor 2 (OLIG2 Antibody)

serum, from guinea pig

Synonym(s):

Oligodendrocyte transcription factor 2, Oligo2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
4
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

guinea pig

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

General description

Oligodendrocyte transcription factor 2 (OLIG2), also known as Oligodendrocyte transcription factor 2 Oligo2, Class B basic helix-loop-helix protein 1 (bHLHb1), Class E basic helix-loop-helix protein 19 (bHLHe19), Protein kinase C-binding protein 2, Protein kinase C-binding protein RACK17, is encoded by the gene name OLIG2/ BHLHB1/ BHLHE19/ PRKCBP2/ RACK17. Olig2 is required for oligodendrocyte and motor neuron specification in the spinal cord, as well as for the development of somatic motor neurons in the hindbrain. It cooperates with OLIG1 to establish the pMN domain of the embryonic neural tube. Olig2 is an antagonist of V2 interneuron and of NKX2-2-induced V3 interneuron development. The restricted expression of Olig2 in oligodendrocytes makes AB9610 an excellent marker of oligodendroglia and oligodendrogliomas in development and mature systems. Olig2 is expressed in the brain and oligodendrocytes. Olig2 is strongly expressed in oligodendrogliomas, while expression is weak to moderate in astrocytomas. Expression in glioblastomas highly variable.
~37 kDa observed

Immunogen

Epitope: Unknown
KLH-conjugated linear peptide corresponding to mouse Oligodendrocyte transcription factor 2 (OLIG2).

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Oligodendrocyte transcription factor 2 (OLIG2) in human cortical brain section tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Oligodendrocyte transcription factor 2 (OLIG2 antibody is validated for use in WB, IH(P) for the detection of Oligodendrocyte transcription factor 2 (OLIG2.

Physical form

Guinea pig polyclonal serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: A 1:1;000 dilution of this antibody detected Oligodendrocyte transcription factor 2 (OLIG2) in 10 µg of mouse brain tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB9610MABN50ABN899
conjugate

unconjugated

conjugate

-

conjugate

-

conjugate

unconjugated

biological source

guinea pig

biological source

rabbit

biological source

mouse

biological source

rabbit

antibody form

serum

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

UniProt accession no.

Q9EQW6

UniProt accession no.

Q13516

UniProt accession no.

Q13516

UniProt accession no.

Q9EQW6

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin), immunoprecipitation (IP): suitable, western blot: suitable

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters