Skip to Content
Merck
All Photos(1)

Key Documents

ABD124

Sigma-Aldrich

Anti-Wnt3a Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein Wnt-3a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WNT3A(89780)
mouse ... Wnt3A(22416)

General description

Wnt-3a, also known as Protein Wnt3a, and encoded by the gene name Wnt3a and Wnt-3a, is a ligand for members of the frizzled family of seven transmembrane receptors and a member of the Wnt family. The Wnt family of consists of ligands involved in cell development, fate determination, organogenesis, and oncogenesis. Information on Wnt signaling has been inferred from genetic analysis of wingless (wg) signaling in Drosophila. The Wnt signaling cascade initiates at the cell membrane when Wnt interacts with its receptor, a member of the Frizzled (Fz) transmembrane receptor family and lipoprotein receptor–related proteins 5 and 6 (LRP-5/6). This interaction relays the signal into the interior of the cell where disheveled (dsh) is brought to the membrane and gets phosphorylated/activated. Dsh promotes the inactivation of GSK3 kinase activity, thus preventing the phosphorylation of β-Catenin and allowing it to accumulate as it avoids ubiquination and subsequent proteosome degradation. Stable β-Catenin accumulates in the cytoplasm where it interacts with lymphoid enhancer factor/T cell factor (LEF/TCF) and translocates into the nucleus. This results in activation of TCF-responsive genes which are thought to play key roles in development and cancer. Negative regulators of the pathway may include axin and the tumor suppressor APC. Wnt-3 and Wnt-3a play distinct roles in cell-cell signaling during morphogenesis of the developing neural tube. Wnt-3a is expressed in dorsal portion of the neural tube (developing roof plate), and mesenchyme tissue surrounding the umbilical veins.

Immunogen

Linear peptide corresponding to mouse Wnt3a.

Application

Immunocytochemistry Analysis: A representative lot detected Wnt3a in COS7 cell lysate expressing WT Wnt3a (Kishida, S., et al. (2004). Mol Cell Biol. 10:4487-501).
Western Blotting Analysis: A representative lot detected Wnt3a in Cos-7 cell lysate overexpressing Wnt-3a (Sato, A., et al. (2010). EMBO J. 29(1):41-54).
Research Category
Stem Cell Research
This Anti-Wnt3a antibody is validated for use in WB, IC for the detection of Wnt3a.

Quality

Evaluated by Western Blotting in Cos 7 overexpressing mouse cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Wnt3a in 10 µg of Cos 7 overexpressing mouse cell lysate.

Target description

~39 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Feng et al.
Molecular medicine reports, 12(3), 4727-4733 (2015-06-23)
Previous studies have demonstrated that the Wnt/β‑catenin signaling pathway is critical to the induction and maintenance of chronic neuropathic pain caused by peripheral inflammation and nerve damage. Emerging evidence from recent studies suggests that epigenetic mechanisms may also be critical
C Hu et al.
Physiological research, 69(4), 701-710 (2020-06-26)
Chemotherapy-induced neuropathic pain (CNP) is the major dose-limiting factor in cancer chemotherapy. However, the mechanisms underlying CNP remain elusive. In the present study, CNP was induced by repeated intraperitoneal injection of vincristine (VCR) into male C57BL/6J mice. VCR administration caused

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service