Skip to Content
Merck
All Photos(2)

Key Documents

ABC847

Sigma-Aldrich

Anti-TACI Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Tumor necrosis factor receptor superfamily member 13B, Transmembrane activator and CAML interactor, CD267

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Members in the TNF superfamily regulate immune responses and induce apoptosis. TACI (also known as Tumor necrosis factor receptor superfamily member 13B, transmembrane activator and CAML interactor, and CD267) binds BAFF and April and its ligands regulate humoral immune responses, activate NF-kB and c-Jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.

Immunogen

Epitope: N-terminus
KLH-conjugated synthetic peptide corresponding to the N-terminus of human TACI.

Application

Detect TACI using this Anti-TACI Antibody validated for use in western blotting & ICC.
Immunocytochemistry Analysis: 2 μg/mL from a representative lot detected TACI in K562 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in K562 and U937 cell lysates.

Western Blotting Analysis: 5 μg/mL of this antibody detected TACI in K562 and U937 cell lysates.

Target description

~32 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service