Skip to Content
Merck
All Photos(2)

Key Documents

ABC464

Sigma-Aldrich

Anti-VAMP-3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Vesicle-associated membrane protein 3, VAMP-3, Cellubrevin, CEB, Synaptobrevin-3

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

General description

VAMP-3 also known as cellubrevin is a vesicular transport protein and SNARE protein important in vesicle trafficking and exocytosis that helps regulate the release of critical components and processes such as blood clotting and inflammatory responses through the release of secretory granules and secretory vesicles. VAMP3 regulates the release of WPBs from endothelial cells. Weibel-Palade bodies (WPBs) are secretory organelles of endothelial cells that store the thrombogenic glycoprotein von Willebrand factor (vWF). Endothelial activation, for example by histamine and thrombin, triggers the Ca(2+)-dependent exocytosis of WPB that releases vWF into the vasculature and thereby initiates platelet capture and thrombus (blood clot) formation. VAMP3 is also critical to the release of proinflammatory compounds IL-6 and TNF-alpha.VAMP-3 is widely expressed in cells.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human VAMP-3 near the N-terminus.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-VAMP-3 Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of VAMP-3.
Western Blotting Analysis: A 1:500 dilution from a representative lot detected VAMP-3 in 10 µg of Huvec and human brain tissue lysate.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected VAMP-3 in PANC-1 cells.

Quality

Evaluated by Western Blotting in PANC-1 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected VAMP-3 in 10 µg of PANC-1 cell lysate.

Target description

~13 kDa observed.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service