Skip to Content
Merck

ABC451

Anti-Dihydropyrimidine dehydrogenase/DPYD Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Dihydropyrimidine dehydrogenase [NADP(+)], DHPDHase, DPD, Dihydrothymine dehydrogenase, Dihydrouracil dehydrogenase, DPYD

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), primate (based on 100% sequence homology), horse (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DPYD(1806)

General description

DPYD is an important enzyme involved in pyrimidine base degradation and catabolism. DPYD catalyzes the reduction of uracil and thymine and also is a critical enzyme in the degradation of the chemotherapeutic drug 5-fluoruracil. DPYD is expressed in most cells and has highest activity in the liver and monocytes. Mutations in DPYD give rise to a range of symptoms and syndromes in patients ranging from no symptoms to highly convulsive and mental retardation caused by excessive amounts of uracil or thymine present in the blood. Because of its ability to degrade 5-fluorouracil, DPYD is studied as a cancer marker and a measure of therapeutic effects in many cancers. Recent research also shows that DPYD levels in the liver are regulated via microRNA interactions and an offer a new opportunity for modulation of this critical enzyme in health and disease.
~110 kDa observed. Uncharacterized band at ~47 kDa may be observed in some lysates.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human Dihydropyrimidine dehydrogenase/DPYD near the N-terminus.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Dihydropyrimidine dehydrogenase/DPYD Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of Dihydropyrimidine dehydrogenase/DPYD.
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected Dihydropyrimidine dehydrogenase/DPYD in 10 µg of human liver tissue lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Dihydropyrimidine dehydrogenase/DPYD in human liver tissue.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human small intestine tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Dihydropyrimidine dehydrogenase/DPYD in 10 µg of human small intestine tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library