Skip to Content
Merck

ABC429

Anti-VEGF-B Antibody

1 mg/mL, from rabbit

Synonym(s):

Vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VEGFB(7423)

General description

VEGF-B, also known as Vascular endothelial growth factor B, VEGF-related factor, VRF, and encoded by the gene VEGFB/VRF, is an endothelial cell growth factor that has multiple splice variants and plays roles in angiogenesis but also in cardiac muscle function, coronary vascular development, chemotaxis of endothelial cells and mast cells, apoptosis regulation, platelet activation, cell division, wound healing and the activation of various signaling pathways. VEGFB is a secreted protein that stays associated with cells because of its ability to bind to heparin. VEGFB is expressed in all cells except for liver cells with the highest levels found in muscle and pancreatic cells. VEGFB levels in plasma are also being studied as a potential biomarker for ventricle damage after heart attacks. High levels of plasma VEGFB show little remodeling whereas patients with lower levels had significant ventricle remodeling, thus VEGFB may have clinical utility in the identification of patients at high risk of remodeling and therefore ventricle damage.
~40/32/25 kDa observed. The ~40kDa band observed is likely due to homo and/or hetero-dimers, the ~32kDa band observed is likely due to glycosylation, and the ~25kDa band is un-modified VEGF186 (Olofsson, B., et al. (1998). Proc Natl Acad Sci USA. 95(20):11709-11714).

Immunogen

Epitope: Unknown
KLH-conjugated linear peptide corresponding to human VEGF-B.

Application

Detect VEGF-B using this Anti-VEGF-B Antibody validated for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected VEGF-B in human skeletal muscle myocytes and vascular tissue and cardiac myocytes in human heart.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human skeletal muscle tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected VEGF-B in 10 µg of human skeletal muscle tissue lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library