Skip to Content
Merck
All Photos(3)

Documents

ABC242

Sigma-Aldrich

Anti-CRKL Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Crk-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CRKL(1399)

General description

Crk-like protein (CRKL) is a 39 kDa adaptor protein with one SH2 and two SH3 binding domains. CRKL is the major protein which is tyrosine phosphorylated in response to BCR/ABL (chronic myelogenous leukemia) and growth factor activation. In myeloid cells, it binds to proteins such as paxillin, p130cas and p120 cbl. CRKL aslo appears to bind ABL and SOS.

Immunogen

KLH-conjugated linear peptide corresponding to human CRKL.

Application

Detect Crk-like protein using this rabbit polyclonal antibody, Anti-CRKL Antibody validated for use in western blotting & IP.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 0.25 µg/mL from a representative lot detected CRKL in 10 µg of NIH3T3 and L6 cell lysate.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated CRKL in 500 µg of Jurkat cell lysate.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected CRKL in 10 µg of Jurkat cell lysate.

Target description

~39 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Ji et al.
Oncology letters, 12(5), 3811-3817 (2016-11-30)
Overexpression of Crk-like (CrkL) adapter protein has been implicated in a number of types of human cancer. However, its involvement in human cervical carcinoma remains unclear. The present study aimed to explore the clinical significance and biological characteristics of CrkL
Le Cai et al.
Oncology letters, 13(1), 51-56 (2017-01-27)
The v-Crk avian sarcoma virus CT10 oncogene homolog-like (CRKL) protein is important in cancer progression. However, its expression pattern and biological roles in human endometrial carcinoma remain unexplored. The potential mechanism of CRKL-induced cancer progression is still unclear. The present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service