Skip to Content
Merck
All Photos(2)

Documents

ABC1486

Sigma-Aldrich

Anti-RIPK1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Receptor-interacting serine/threonine-protein kinase 1, RIPK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ripk1(19766)

General description

Receptor-interacting serine/threonine-protein kinase 1, also known as Cell death protein RIP or Receptor-interacting protein 1 (RIP-1) or Serine/threonine-protein kinase RIP, and encoded by the gene name RIPK1 or RIP or RIP1, contains 1 death domain and 1 protein kinase domain. RIPK1 is localized to the cytoplasm and plays an important role in a variety of cellular pathways including the NF-κB pathway and programmed necrotic cell death (necroptosis).

Immunogen

Epitope: Near N-terminus
RIPK1 antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human RIPK1.

Application

Immunohistochemistry Analysis (IHC): 2.5 μg/mL from a representative lot detected RIPK1 in mouse kidney tissue.

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-RIPK1 Antibody is validated for use in Western Blotting_x000D_ and Immunohistochemistry for the detection of RIPK1.

Quality

Evaluated by Western Blotting in rat kidney tissue lysate.

Western Blotting Analysis (WB): 1 μg/mL of this antibody detected RIPK1 in rat kidney tissue lysate.

Target description

~75 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service