Skip to Content
Merck

Skip To

ABC129

Anti-WWP2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

NEDD4-like E3 ubiquitin-protein ligase WWP2, WW domain-containing protein 2

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
1


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WWP2(11060)

General description

WW2 (atrophin-1-interacting protein 2) is a member of a large family of NEDD4-like E3 ligases that transfer ubiquitin from E2 ubiquitin-conjugating enzymes to targeted protein substrates. In general, ubiquitination affects protein turnover and activity. As a result, ubiquitin-related enzymes play essential roles in many cellular processes. The WW2 E3 ubiquitin-protein ligase contains a C2 domain at the N-terminus and a HECT (E6AP-type E3 ubiquitin-protein ligase) domain at the C-terminus which interact to inhibit its catalytic activity. Previous studies have suggested that WW2 may play a role in cellular transport, transcription, and T-cell activation. It has also been shown that WW2 regulates the PTEN tumor suppressor and plays an essential role in apoptosis and survival of cancer cells. WW2-deficient mice show abnormal development of craniofacial structures.
~100 kDa observed. Uncharacterized bands may be observed at ~31 kDa, ~57 kDa and ~60 kDa in some cell lysates.

Immunogen

GST-tagged recombinant protein corresponding to mouse WWP2.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-WWP2 Antibody is validated for use in Western Blotting, IHC for the detection of WWP2.
Western Blot Analysis: A representative lot of this antibody detected WWP2 in CGR8 ES whole cell lysate and in transiently transfected HEK293 cells (Xu, H. M. et al. (2004). J Biol Chem. 279(22):23495-23503.).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
mESC lysate
Evaluated by Western Blot in mESC lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected WWP2 in 10 µg of mESC lysate (For optimal performance, a range of 1 to 2 µg/mL of this antibody is recommended in Western Blot).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABE473AB4350MABF1110
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

Gene Information

human ... WWP2(11060)

Gene Information

human ... RNF20(56254)

Gene Information

human ... PRDM14(63978)

Gene Information

human ... UBR5(51366)

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

ambient


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
ABC12904053252696657

Questions

Reviews

No rating value

Active Filters