Skip to Content
Merck
All Photos(1)

Documents

AB9554

Sigma-Aldrich

Anti-Capsaicin Receptor Antibody

serum, Chemicon®

Synonym(s):

Vanilloid Receptor, VR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TRPV1(7442)

Specificity

Recognizes Capsaicin Receptor (VR1, Vallinoid Receptor 1, TRPV1).

Immunogen

Synthetic peptide from the C-terminal of mouse VR1.

Application

Anti-Capsaicin Receptor Antibody is an antibody against Capsaicin Receptor for use in IH.
Immunohistochemistry: 1:500-1:1,000 on mouse DRG tissued fixed with 2% paraformaldehyde.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Neurotransmitters & Receptors

Physical form

Rabbit serum. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah A Stanley et al.
Nature medicine, 21(1), 92-98 (2014-12-17)
Means for temporally regulating gene expression and cellular activity are invaluable for elucidating underlying physiological processes and would have therapeutic implications. Here we report the development of a genetically encoded system for remote regulation of gene expression by low-frequency radio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service