Skip to Content
Merck
All Photos(1)

Documents

AB5884

Sigma-Aldrich

Anti-HCN1 Antibody

Chemicon®, from rabbit

Synonym(s):

Hyperpolarization-Activated Cyclic Nucleotide-Gated Potassium Channel 1, HAC2, BCNG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HCN1(348980)

Specificity

Recognizes HCN-1 (Hyperpolarization-Activated Cyclic Nucleotide-Gated Potassium Channel 1, HAC2, BCNG1). The epitope does not share homology with any other known proteins.

SPECIES REACTIVITIES: It is expected that the antibody will also react with mouse (18/19) and possibly rabbit (16/19) due to sequence homology. Other species have not been tested.

Immunogen

Synthetic peptide corresponding amino acids 6-24 of rat HCN-1 (Accession number AAF62173).

Application

Anti-HCN1 Antibody is an antibody against HCN1 for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1:200-1:400 using ECL on rat brain membranes.

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 40 μg of control antigen. The stock solution of the antigen can be made up using 100 μL of sterile distilled water. For negative control, preincubate 1 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuhki Saito et al.
Molecular neurodegeneration, 7, 50-50 (2012-10-05)
One of the best-characterized causative factors of Alzheimer's disease (AD) is the generation of amyloid-β peptide (Aβ). AD subjects are at high risk of epileptic seizures accompanied by aberrant neuronal excitability, which in itself enhances Aβ generation. However, the molecular
Shui-Wang Ying et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(28), 10412-10423 (2011-07-15)
Hyperpolarization-activated cyclic nucleotide-gated (HCN) channels generate a pacemaking current, I(h), which regulates neuronal excitability and oscillatory activity in the brain. Although all four HCN isoforms are expressed in the brain, the functional contribution of HCN3 is unknown. Using immunohistochemistry, confocal
Damien Le Menuet et al.
Cardiovascular research, 87(3), 467-475 (2010-03-20)
Cardiac mineralocorticoid receptor (MR) activation triggers adverse cardiovascular events that could be efficiently prevented by mineralocorticoid antagonists. To gain insights into the pathophysiological role of MR function, we established embryonic stem (ES) cell lines from blastocysts of transgenic mice overexpressing
Choong-Hyun Lee et al.
Iranian journal of basic medical sciences, 22(11), 1308-1313 (2020-03-05)
Hyperpolarization-activated cyclic nucleotide-gated (HCN) channels play essential roles in various hippocampal functions, including regulation of long-term potentiation, synaptic plasticity, and hippocampal-dependent cognitive process. The objective of this study was to investigate age-related changes in HCN1 and HCN2 protein expressions in
Regulation of the hyperpolarization-activated cationic current Ih in mouse hippocampal pyramidal neurones by vitronectin, a component of extracellular matrix
Vasilyev, Dmitry V and Barish, Michael E
The Journal of Physiology, 560, 659-675 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service