Skip to Content
Merck
All Photos(1)

Key Documents

AB5512

Sigma-Aldrich

Anti-Homocysteine Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ELISA
species reactivity:
rat
technique(s):
ELISA: suitable
citations:
2

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

shipped in

dry ice

target post-translational modification

unmodified

General description

Homocysteine a sulphur containing amino acid, is formed by the transmethylation of methionine, an essential amino acid. Homocysteine may be irreversibly metabolised to cystathionine by cystathionine synthetase. This reaction uses pyridoxal phosphate (Vitamin B6) as a co-enzyme (see figure). The enzyme cystathionine synthetase is deficient in homocysteinuria, a rare genetic disease associated with high blood levels of homocysteine and the development of premature atherosclerosis. This and the association of other rare enzyme deficiencies with homocysteinemia and premature atherosclerosis led McCully to formulate the homocysteine hypothesis of atherosclerosis (McCully KS, The homocysteine theory of atherosclerosis. Atherosclerosis. 1976; 20: 23-27)

Specificity

Homocysteine



The cross-reactivities were determined using an ELISA test by competition experiments with the following compounds:



Compound Cross-reactivity ratio

Homocysteine-G-BSA 1

Cysteine-G-BSA 1/450

Homocysteic acid-G-BSA 1/>100,000



Abbreviations:



(G) Glutaraldehyde

(BSA) Bovine Serum Albumin

Immunogen

Homocysteine-Glutaraldehyde-BSA

Application

Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism

Neurodegenerative Diseases
Suggested working dilution: 1:1,000-1:5,000

Optimal working dilutions must be determined by the end user.
This Anti-Homocysteine Antibody is validated for use in ELISA for the detection of Homocysteine.

Physical form

Rabbit antiserum. Liquid with 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amany Tawfik et al.
Journal of clinical medicine, 8(1) (2019-01-24)
Diabetic retinopathy (DR) is the most common cause of blindness in people under the age of 65. Unfortunately, the current screening process for DR restricts the population that can be evaluated and the disease goes undetected until irreversible damage occurs.
Hieronim Jakubowski
Physiological reviews, 99(1), 555-604 (2018-11-15)
Epidemiological studies established that elevated homocysteine, an important intermediate in folate, vitamin B12, and one carbon metabolism, is associated with poor health, including heart and brain diseases. Earlier studies show that patients with severe hyperhomocysteinemia, first identified in the 1960s

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service