Skip to Content
Merck
All Photos(1)

Documents

AB5454

Sigma-Aldrich

Anti-TUC-4 Protein Antibody

serum, Chemicon®

Synonym(s):

ULIP-1 Protein, CRMP-4, DRP-3, Dihydropyrimidinase-like 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

feline, monkey, human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DPYSL3(1809)

General description

TUC (TOAD [Turned On After Division]/ Ulip/CRMP) is a family of proteins that have emerged as strong candidates for a role in neuronal growth cone signaling. The TUC-4 family members reach their highest expression levels in early post-mitotic neurons during their peak periods of axonal growth. TUC-4 is not expressed in neural progenitors or adult neurons (with minor exception). Data suggests that TUC-4 plays a role in axonal outgrowth, pathfinding and possibly in the process that permits growing axons to choose proper routes and targets (Quinn, 1999). The TUC-4 protein is expressed primarily within the cytoplasm of postmitotic neurons as they begin their migration out of the ventricular zone into the developing cortical plate (Minturn, 1995a). TUC-4 is expressed in neuronal cell bodies, neurites, and along the most distal regions of neuronal growth cones, including the lamellipodia and filopodia (Minturn, 1995b). Expression of TUC-4 is observed by day E12 in rats coincident with the expression of the early neuronal marker class III beta-tubulin (Quinn, 1999). TUC-4 expression is rarely observed in neuronal cells expressing the mitotic marker PCNA (Minturn, 1995a). The expression of TUC-4 has been shown to be upregulated in response to NGF in rats (Minturn, 1995b) and in response to retinoic acid in a human neuroblastoma cell line (SMS-KCNR; Gaetano, 1997).

Specificity

Specifically recognizes the TUC-4 (Ulip-1/CRMP-4/DRP-3) splice variants, TUC-4a (64 kDa) and the N-terminally extended TUC-4b (75 kDa) (Quinn, 2003). AB5454 has been reported to label early postmitotic neurons in prenatal rat brain. Labeling is observed primarily within the cytoplasm along the most distal regions of neuronal growth cones including lamellipodia and filopodia. The antibody is not reactive with the proteins TUC-1 (Ulip-3/CRMP-1), TUC-2 (Ulip-2/CRMP-2) or TUC-3 (Ulip-4/CRMP-3). It does not label neural progenitors or radial glia.

Immunogen

Synthetic peptide corresponding to amino acids 499 to 511 of TUC-4a and amino acids 612-624 of TUC-4b (YDGPVFDLTTTPK)

Application

Anti-TUC-4 Protein Antibody detects level of TUC-4 Protein & has been published & validated for use in IC, IH, IP & WB.
Immunohistochemistry: 1:1,000-1:5,000 using PFA (DAB detection).
Immunocytochemistry: 1:1,000-1:5,000 (DAB detection).
Western blot: 1:2,500-1:25,000 using alkaline phosphatase.
Immunoprecipitation

Optimal working dilutions must be determined by end user.

Target description

64 & 74 kDa

Physical form

Rabbit Serum. Liquid containing 0.2% sodium azide.

Analysis Note

Control
POSITIVE CONTROL: Fetal brain/spinal cord tissue. Human neuroblastoma cell line SMS-KCNR or rat PC12 cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kunlin Jin et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 24(4), 399-408 (2004-04-17)
Heparin-binding epidermal growth factor-like growth factor (HB-EGF) is a hypoxia-inducible, neuroprotective protein that also stimulates proliferation of neuronal precursor cells. Accordingly, HB-EGF may contribute to recovery from cerebral injury through direct neuroprotective effects, by enhancing neurogenesis, or both. When administered
Yulia Vainshenker et al.
Clinical medicine insights. Case reports, 10, 1179547617732040-1179547617732040 (2017-10-06)
Patient recovering from traumatic vegetative state has suddenly died from cardiac arrest. In-life improvement of consciousness appeared after reduction of generalized spasticity due to botulinum toxin administration. Neuropathologic examination revealed Musashi1+, Nestin+, PCNA+, and Ki67+ cells in the hippocampus, frontal
Patrick Gelé et al.
Proteome science, 12, 24-24 (2014-06-20)
Lipid lowering agent such as agonists of peroxisome proliferator-activated receptors (PPAR) are suggested as neuroprotective agents and may protect from the sequelae of brain ischemic stroke. Although the demonstration is not clearly established in human, the underlying molecular mechanism may
Lori B Bennett et al.
Neuroscience letters, 475(1), 1-6 (2010-03-20)
Production of new neurons throughout adulthood has been well characterized in two brain regions, the subventricular zone (SVZ) of the anterolateral ventricle and the subgranular zone (SGZ) of the hippocampus. The neurons produced from these regions arise from neural stem
Jun Nagai et al.
Scientific reports, 5, 8269-8269 (2015-02-06)
Axonal outgrowth inhibitors and scar formation are two major obstacles to central nervous system (CNS) repair. No target molecule that regulates both axonal growth and scarring has been identified. Here we identified collapsin response mediator protein 4 (CRMP4), a common

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service