Skip to Content
Merck
All Photos(2)

Key Documents

AB5320B

Sigma-Aldrich

Anti-NG2 Chondroitin Sulfate Proteoglycan, Biotin Conjugate Antibody

from rabbit, biotin conjugate

Synonym(s):

Chondroitin sulfate proteoglycan 4, Chondroitin sulfate proteoglycan NG2, Melanoma chondroitin sulfate proteoglycan, Melanoma-associated chondroitin sulfate proteoglycan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

biotin conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), monkey (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CSPG4(1464)
mouse ... Cspg4(121021)
rat ... Cspg4(81651)
rhesus monkey ... Cspg4(713086)

General description

NG2 is a high molecular weight, integral membrane chondroitin sulfate proteoglycan. It is found on the surfaces of an unusual class of glial cells within the developing and mature central nervous system that have the properties of oligodendrocyte precursor cells (i.e., O-2A progenitor cells). NG2 is also found on the surfaces of chondroblasts, proliferating capillary endothelial cells, some human melanoma cell lines, and on leukemic blasts in childhood acute lymphoblastic leukemia. The NG2 proteoglycan is likely to play a role in regulation of cell motility, axon outgrowth and the cellular responses to platelet-derived growth factor.

Application

Anti-NG2 Chondroitin Sulfate Proteoglycan, Biotin Conjugate Antibody is an antibody against NG2 Chondroitin Sulfate Proteoglycan for use in IHC, ICC.
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected NG2 in paraffin embedded rat cortex tissue.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected NG2 in rat oligodendrocyte precursor cells. Performed without Triton.

Quality

Evaluated by Immunohistochemistry in cryosections of rat cortex tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected NG2 in cryosections of rat cortex tissue.

Target description

The unconjugated parent antibody (Cat. No. AB5320) has an observed molecular weight at ~270-300 kDa

Analysis Note

Control
Cryosections of rat cortex tissue

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meera Murgai et al.
Nature medicine, 23(10), 1176-1190 (2017-09-19)
A deeper understanding of the metastatic process is required for the development of new therapies that improve patient survival. Metastatic tumor cell growth and survival in distant organs is facilitated by the formation of a pre-metastatic niche that is composed
Sonja Rittchen et al.
Biomaterials, 56, 78-85 (2015-05-03)
Multiple sclerosis (MS) is a progressive demyelinating disease of the central nervous system (CNS). Many nerve axons are insulated by a myelin sheath and their demyelination not only prevents saltatory electrical signal conduction along the axons but also removes their
Deyu Hu et al.
Stem cell research & therapy, 15(1), 199-199 (2024-07-07)
Bone marrow-derived mesenchymal stem cell (BMMSC)-based therapy has become a major focus for treating liver fibrosis/cirrhosis. However, although these cell therapies promote the treatment of this disease, the heterogeneity of BMMSCs, which causes insufficient efficacy during clinical trials, has not
Alexander Balatskiy et al.
International journal of molecular sciences, 23(3) (2022-02-16)
The local development of atherosclerotic lesions may, at least partly, be associated with the specific cellular composition of atherosclerosis-prone regions. Previously, it was demonstrated that a small population of immature vascular smooth muscle cells (VSMCs) expressing both CD146 and neuron-glial
Qionghong Xie et al.
Cell death & disease, 13(12), 1044-1044 (2022-12-16)
Accumulating evidence indicates that the extracellular matrix (ECM) is not only a consequence of fibrosis, but also contributes to the progression of fibrosis, by creating a profibrotic microenvironment. Tenascin-C (TNC) is an ECM glycoprotein that contains multiple functional domains. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service