Skip to Content
Merck
All Photos(3)

Key Documents

AB356482

Sigma-Aldrich

Anti-mCherry

Anti-mCherry

from rabbit

Synonym(s):

Anti-mCherry Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

all

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

mCherry, red fluorescent protein, is derived from DsRed, a red fluorescent protein from disc corals of Discosoma species. DsRed is similar in size and other properties to GFP, but it produces a red rather than a green fluorescence. mCherry is a monomeric form that has been engineered with multiple cycles of mutation, directed modification and evolutionary selection. It exhibits excitation maximum at 587 nm and an emission maximum at 610 nm with a fluorescent quantum yield of 0.22. mCherry displays high photostability and resistant to photobleaching. It also displays improved brightness and extremely rapid maturation rate. Anti-mCherry, polyclonal is an affinity purified antibody that recognizes mCherry in HEK293 cells transfected with pFin-EF1-mCherry. (Ref.: Schaner, NC et al. (2004). Nat. Biotechnol. 22(12):1567-72; Gross, LA et al. (2000). Proc. Natl. Acad. Sci. USA. 97(22):11990-95).

Specificity

This polyclonal antibody detects mCherry in HEK293 cells tranfected with pFin-mCherry.

Immunogen

Epitope: unknown
His-tagged full length recombinant mCherry from Discosoma sp (Mushroom anemone).

Application

Detect mCherry protein using this rabbit polyclonal anti-mCherry Antibody, Cat. No. AB356482. It has been tested in Immunofluorescence and Western Blotting.
Immunofluorescence Analysis: A 1:500 dilution from a representative lot detected mCherry in HEK293 cells transfected with pFin-EF1-mCherry vector. (Courtesy of Dr Gerry Shaw).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected mCherry in HEK293 cells transfected with pFin-EF1-mCherry vector. (Courtesy of Dr Gerry Shaw).
Research Category
Secondary & Control Antibodies

Quality

Evaluated by Western Blotting in HEK293 cells transfected with pFin-EF1-mCherry vector.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected mCherry in 10 µg lysate from HEK293 cells transfected with pFin-EF1-mCherry vector.

Target description

~32 kDa observed, 26.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 0.03% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao-Shan Chen et al.
Nature communications, 13(1), 3605-3605 (2022-06-24)
The cell-type-specific recording and manipulation is instrumental to disentangle causal neural mechanisms in physiology and behavior and increasingly requires intersectional control; however, current approaches are largely limited by the number of intersectional features, incompatibility of common effectors and insufficient gene
Vijeta Jaiswal et al.
Frontiers in bioengineering and biotechnology, 11, 1142415-1142415 (2023-05-08)
Aedes mosquitoes transmit several pathogens including flaviviruses to humans which result in high morbidity and mortality. Owing to adaptability and climate change, these mosquito vectors are predicted to establish in new geographical areas thus exposing larger populations to the risk
Tong Xiao et al.
Nature chemical biology, 14(7), 655-663 (2018-06-06)
The unusually high demand for metals in the brain, along with insufficient understanding of how their dysregulation contributes to neurological diseases, motivates the study of how inorganic chemistry influences neural circuitry. We now report that the transition metal copper is
Gen Li et al.
EMBO reports, 22(3), e51163-e51163 (2021-01-26)
The phagosome harboring the bacterial pathogen Legionella pneumophila is known to be enriched with phosphatidylinositol 4-phosphate (PtdIns4P), which is important for anchoring a subset of its virulence factors and potentially for signaling events implicated in the biogenesis of the Legionella-containing
Marina Capece et al.
Cell death and differentiation, 30(9), 2078-2091 (2023-08-04)
The discrimination of protein biological functions in different phases of the cell cycle is limited by the lack of experimental approaches that do not require pre-treatment with compounds affecting the cell cycle progression. Therefore, potential cycle-specific biological functions of a

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service