Skip to Content
Merck
All Photos(1)

Key Documents

AB3276

Sigma-Aldrich

Anti-Aquaporin 3 Antibody

Chemicon®, from rabbit

Synonym(s):

AQP3, GLIP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
clone:
polyclonal
application:
IHC
WB
species reactivity:
rat, mouse
technique(s):
immunohistochemistry: suitable
western blot: suitable
citations:
8

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AQP3(360)

Specificity

Recognizes Aquaporin 3.

SPECIES REACTIVITIES: The epitope specific for Aquaporin 3 is highly homologous in human and sheep (15/18). Reactivity with other species has not yet been tested.

Immunogen

Highly purified peptide corresponding to residues 275-292 of rat Aquaporin 3 (Accession Number P47862) coupled to KLH.

Application

Anti-Aquaporin 3 Antibody detects level of Aquaporin 3 & has been published & validated for use in WB, IH.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blotting: 1:200 on rat kidney membranes. Identifies a 31kDa and 40kDa band. The large band may represent the glycosylated form of the protein.

Immunohistochemistry on mouse kidney sections.

Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, 5% sucrose and 0.025% sodium azide. Reconstitute with 200 μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 6 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). Reconstitute with 100 μL of deionized water. For negative control, preincubate 1 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Nagai et al.
Neurourology and urodynamics, 38(6), 1560-1570 (2019-06-14)
To measure the effects of nicotine on lower urinary tract symptoms (LUTS), bladder blood flow, and the urothelial markers hypoxia-inducible factor 1α (HIF1α), uroplakin III (UPIII), and aquaporin 3 (AQP3). Ten-week-old female Sprague Dawley rats were subcutaneously injected with 2 mg/kg
Qi Chen et al.
Cell research, 21(6), 922-933 (2010-12-08)
In the journey from the male to female reproductive tract, mammalian sperm experience a natural osmotic decrease (e.g., in mouse, from ~415 mOsm in the cauda epididymis to ~310 mOsm in the uterine cavity). Sperm have evolved to utilize this
Jae Mi Suh et al.
Molecular endocrinology (Baltimore, Md.), 20(12), 3412-3420 (2006-09-16)
Nuclear receptors are transcriptional regulators that play important roles in embryonic development and organogenesis. To study the potential roles of nuclear receptors in kidney development, we examined the expression patterns of a subset of nuclear receptors in which specific antibodies
Harumi Hisaki et al.
American journal of physiology. Renal physiology, 303(7), F1049-F1059 (2012-07-27)
Saposin D-deficient (Sap-D(-/-)) mice develop polydipsia/polyuria and die prematurely due to renal failure with robust hydronephrosis. Such symptoms emerged when they were around 3 mo of age. To investigate the pathogenesis of their water mishandling, we attempted to limit water
Kyungmi Yang et al.
Marine drugs, 18(8) (2020-07-28)
Radiation dermatitis (RD) is one of the most common side effects of radiotherapy; its symptoms progress from erythema to dry and moist desquamation, leading to the deterioration of the patients' quality of life. Active metabolites in brown seaweed, including phlorotannins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service