Skip to Content
Merck
All Photos(1)

Key Documents

AB3242

Sigma-Aldrich

Anti-PGC-1 Antibody

Chemicon®, from rabbit

Synonym(s):

Peroxisome proliferator-activated receptor gamma coactivator 1-alpha, Ligand effect modulator 6, PGC-1-alpha, PPAR-gamma coactivator 1-alpha, PPARGC-1-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

PGC-1 is a transcription factor that determines whether preadipocytes will differentiate into brown or white fat cells. It interacts with PPARgamma to promote the expression of a cassette of genes which define the brown adipocyte phenotype.

Specificity

Immunogen peptide blocking prior to Western blotting completely abolished target band detection by this rabbit polyclonal antibody.

Immunogen

Epitope: C-terminus
Synthetic peptide from amino acids 777-797 (C-terminus) of mouse PGC-1.

Application

Anti-PGC-1 Antibody is a rabbit olyclponal antibody for the detection of PGC-1 also known as PPARgamma Coactivator 1-alpha & has been validated in ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
ELISA Analysis: A 1:75,000 dilution of a previous lot was used in ELISA.

Immunocytochemistry Analysis: A 1:300 dilution of a previous lot was used in Immunocytochemistry.

Immunofluorescence Analysis: Representative lots detected PGC-1α in frozen mouse and rat muscle tissue sections (Gouspillou, G., et al. (2013). Longev. Healthspan. 2(1):13; Matsuura, T., et al. (2007). J. Orthop. Res. 25(11):1534-1540).

Immunohistochemistry Analysis: A representative lot detected PGC-1α in free-floating mouse forebrain sections (Tallaksen-Greene, S.J., et al. (2014). J Neurosci. 34(47):15658-15668).

Immunoprecipitation Analysis: Representative lots immunoprecipitated PGC-1α from mouse liver, skeletal muscle, and brown adipose tissue lysates (Boutant, M., et al. (2016). Cell Rep. 14(9):2068-2075; Park, R.D., et al. (2014). J. Exerc. Nutrition Biochem. 18(1):1-7; Kim, J.S., et al. (2014). J. Exerc. Nutrition Biochem. 18(3):259-266; Philip, A., et al. (2011). J. Biol. Chem. 286(35):30561-30570).

Western Blotting Analysis: Representative lots detected similar PGC-1α in human, mouse, and rat skeletal muscle tissue, as well as in nuclear and mitochondrial preparations from rat brown adipose tissue (Suwa, M., et al. (2015). J. Sports Sci. Med. 14(3):548-555; Lee, I., et al. (2015). Front. Pharmacol. 6:43; Vincent, G., et al. (2015). Front Physiol. 6:51; Lombardi, A., et al. (2015). PLoS One. 10(2):e0116498; Gouspillou, G., et al. (2014). FASEB J. 28(4):1621-1633; Saleem, A., et al. (2014). Am. J. Physiol. Cell Physiol. 306(3):C241-249).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in mouse BAT extracts.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected recombinant mouse PGC-1, as well as endogenous PGC-1 in 15 µg of nuclear extract and whole cell lysate from mouse brown adipose tissue (BAT). Immunogen peptide blocking completely blocked target band detection.

Target description

~100 kDa observed. 91.03/89.64/91.71/77.14 kDa (human isoform 1/B4/B5/L-PGG-1alpha), 90.59 kDa (mouse isoform 1; PGC-1a1), 90.62 kDa (rat) calculated.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 20 mM PBS and 0.1% sodium azide.

Storage and Stability

Stable for 1 year at -70ºC from date of receipt in undiluted aliquots. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
BAT nuclear prep

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong-Kook Lee et al.
Scientific reports, 5, 16364-16364 (2015-11-07)
Obesity is often associated with irregular dietary habits and reduced physical activity. Regular exercise induces a metabolic response that includes increased expression of various cytokines, signaling proteins and hormones, and reduced adipocyte size. In this study, mice performed a swimming
High fat diet induced developmental defects in the mouse: oocyte meiotic aneuploidy and fetal growth retardation/brain defects.
Luzzo, KM; Wang, Q; Purcell, SH; Chi, M; Jimenez, PT; Grindler, N; Schedl, T; Moley, KH
Testing null
Exercise training improves plantar flexor muscle function in mdx mice.
Baltgalvis, KA; Call, JA; Cochrane, GD; Laker, RC; Yan, Z; Lowe, DA
Med Sci Sports Exerc. null
Role of peroxisome proliferator-activated receptor gamma coactivator 1-alpha (PGC-1?) in denervation-induced atrophy in aged muscle: facts and hypotheses.
Gouspillou, G; Picard, M; Godin, R; Burelle, Y; Hepple, RT
Longevity & healthspan null
Divergent skeletal muscle respiratory capacities in rats artificially selected for high and low running ability: a role for Nor1?
Stephenson, EJ; Stepto, NK; Koch, LG; Britton, SL; Hawley, JA
Journal of Applied Physiology (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service