Skip to Content
Merck
All Photos(1)

Key Documents

AB2031

Sigma-Aldrich

Anti-Collagen Type II Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Col2A1(12824)

Specificity

Antibody shows by RIA cross reactivity with mouse collagen types I, III, IV of less than 0.1% at a dilution of 1:100. The antibody also demonstrates a reactivity with mouse fibronectin of less than 0.1% by RIA at dilution of 1:100. There is also cross reactivity with bovine collagen type II by ELISA

Immunogen

Collagen Type II extracted and purified from mouse fetal cartilage.

Application

Anti-Collagen Type II Antibody is an antibody against Collagen Type II for use in ELISA, IF, RIA, IH.
Immunohistochemistry: 1:40-1:80 dilution for immunofluorescent staining of fresh frozen mouse articular tissue. Acetone or methyl-carnoy fixation is also reactive.

Radioimmunoassay

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Protein G affinity purified IgG fraction at 1.0 mg/mL in 0.01M phosphate, 0.09M NaCl, pH 7.2 with no preservatives.

Storage and Stability

Maintain frozen at -20°C for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Important Note: During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yucheng Lin et al.
Arthritis research & therapy, 26(1), 111-111 (2024-05-30)
Due to the unclear pathogenesis of osteoarthritis (OA), effective treatment for this ailment is presently unavailable. Accumulating evidence points to chondrocyte senescence as a key driver in OA development. This study aims to identify OA-specific microRNAs (miRNAs) targeting chondrocyte senescence
Bone mesenchymal stem cell-derived extracellular vesicles promote the repair of intervertebral disc degeneration by transferring microRNA-199a.
Wen, et al.
Cell Cycle, 20, 256-270 (2022)
Issaka Yougbaré et al.
The Journal of clinical investigation, 125(4), 1545-1556 (2015-03-17)
Fetal and neonatal alloimmune thrombocytopenia (FNAIT) is a life-threatening disease in which intracranial hemorrhage (ICH) is the major risk. Although thrombocytopenia, which is caused by maternal antibodies against β3 integrin and occasionally by maternal antibodies against other platelet antigens, such
Ivana Gadjanski et al.
Tissue engineering. Part A, 19(19-20), 2188-2200 (2013-05-09)
Formation of tissue-engineered cartilage is greatly enhanced by mechanical stimulation. However, direct mechanical stimulation is not always a suitable method, and the utilization of mechanisms underlying mechanotransduction might allow for a highly effective and less aggressive alternate means of stimulation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service