Skip to Content
Merck
All Photos(1)

Key Documents

AB1576

Sigma-Aldrich

Anti-Cysteine String Protein Antibody

serum, Chemicon®

Synonym(s):

CSP, DnaJ Homologue Subfamily C member 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DNAJC5(80331)

General description

Cysteine-string proteins are membrane-bound proteins associated primarily with nerve endings and synaptic vesicles in the brain where they serve an important role in presynaptic function and may be involved in calcium-dependent neurotransmitter release at nerve endings. CSPs posses a strech of amino acids highly enriched in cysteine residues that are post-translationally palmitoylated. CSPs may function as homodimers.

Specificity

Cysteine-string Protein. The antibody recognizes primarily the alpha isoform of CSP with some recognition of the beta isoforms. Slight reactivity with the gamma isoforms may be detected.

Immunogen

Recombinant rat cysteine-string protein with the cysteine string deleted.

Application

Immunohistochemistry on formalin fixed tissue: 1:1,000. (Recommend Cat. No. AB1576P).Immunocytochemistry on PC-12 cells: 1:1,000. (Recommend Cat. No. AB1576P).Immunoblotting: 1:5,000. CSPs should be resolved on 12-15% SDS-PAGE gels under NON-REDUCING conditions. CSPs typically migrate as 33-35 kDa species in vertebrate samples (dimers may also be seen). If one must perform SDS-PAGE under reducing conditions, it is necessary to alkylate the CSPs to prevent smearing.To ensure proper identification of CSPs, samples may be deacylated by treatment with 1M neutral hydroxylamine or 0.1M KOH in methanol resulting in a 6-7 kDa shift in electrophoretic mobility of the CSP band. Deacylation will usually result imn some loss of reactivity on western blots so sufficient protein should be loaded. Alternatively, because all CSPs appear to be acidic (approximate pI of 4.6-4.8) 2-D gel electrophoresis may be usefull to distinguish some CSPs. Finally, some sample treatments may cause partial deacylation of CSPs resulting in a smear of immunoreactivity below the mass of the fatty acylated monomer.Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
This Anti-Cysteine String Protein Antibody is validated for use in WB, IC, IH for the detection of Cysteine String Protein.

Physical form

Liquid rabbit serum containing no preservatives

Storage and Stability

Store at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: brain samples

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huzefa Rupawala et al.
Brain communications, 4(4), fcac192-fcac192 (2022-08-06)
In Alzheimer's disease, synapse loss causes memory and cognitive impairment. However, the mechanisms underlying synaptic degeneration in Alzheimer's disease are not well understood. In the hippocampus, alterations in the level of cysteine string protein alpha, a molecular co-chaperone at the
Laura Caló et al.
Brain : a journal of neurology, 144(6), 1661-1669 (2021-03-25)
α-Synuclein aggregation at the synapse is an early event in Parkinson's disease and is associated with impaired striatal synaptic function and dopaminergic neuronal death. The cysteine string protein (CSPα) and α-synuclein have partially overlapping roles in maintaining synaptic function and
Evidence that the presynaptic vesicle protein CSPalpha is a key player in synaptic degeneration and protection in Alzheimer's disease.
Tiwari, SS; d'Orange, M; Troakes, C; Shurovi, BN; Engmann, O; Noble, W; Hortobagyi, T; Giese, KP
Molecular Brain null
Lukas Rüttiger et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(35), 12922-12927 (2004-08-26)
The large conductance voltage- and Ca2+-activated potassium (BK) channel has been suggested to play an important role in the signal transduction process of cochlear inner hair cells. BK channels have been shown to be composed of the pore-forming alpha-subunit coexpressed
Ching-Feng Chen et al.
Visual neuroscience, 39, E003-E003 (2022-05-12)
During the first postnatal week in rodents, cholinergic retinal waves initiate in starburst amacrine cells (SACs), propagating to retinal ganglion cells (RGCs) and visual centers, essential for visual circuit refinement. By modulating exocytosis in SACs, dynamic changes in the protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service