Skip to Content
Merck
All Photos(1)

Documents

AB1572

Sigma-Aldrich

Anti-GABA Transporter-2 Antibody

Chemicon®, from rabbit

Synonym(s):

GAT-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

GABA Transporter-2 (GAT-2), C-terminus. No cross-reactivity to the C-termini of other transmitter transporters detected.

Immunogen

GAT-2, C-terminus (aa 593-602)

Application

Immunohistochemistry: 1:500-1:750 on 4% paraformaldehyde fixed tissue.

It is recommended that the antibody is diluted in PBS containing 10% normal goat serum and 0.5% Triton X-100. AB1572 has been used for light microscopic studies of the rat nervous system.

Western blot: It has been reported that this antibody works for Western blot (Conti, 1999).

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-GABA Transporter-2 Antibody is validated for use in IH, WB for the detection of GABA Transporter-2.

Physical form

Affinity purified antibody. Purified in PBS with 0.1% sodium azide.

Storage and Stability

Maintain lyophilized material at 2-8°C for up to 12 months. Maintain reconstituted material at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Lin et al.
Disease models & mechanisms, 10(12), 1529-1538 (2017-12-21)
Friedreich ataxia (FRDA) is an autosomal recessive neurodegenerative disorder with progressive ataxia that affects both the peripheral and central nervous system (CNS). While later CNS neuropathology involves loss of large principal neurons and glutamatergic and GABAergic synaptic terminals in the
Amanda L Smith et al.
Scientific reports, 10(1), 7834-7834 (2020-05-14)
Inhibitory interneurons are essential for proper brain development and function. Dysfunction of interneurons is implicated in several neurodevelopmental disorders, including autism spectrum disorder (ASD) and intellectual disability (ID). We have previously shown that Arid1b haploinsufficiency interferes with interneuron development and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service