Skip to Content
Merck
All Photos(1)

Key Documents

AB15688

Sigma-Aldrich

Anti-Adrenergic β3 receptor Antibody

Chemicon®, from chicken

Synonym(s):

ADRB3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

chicken

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ADRB3(155)

Specificity

Adrenergic beta3 receptor (ADRB3).

Immunogen

Synthetic peptide from mouse ADRB3.

Application

Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-Adrenergic β3 receptor Antibody is validated for use in WB for the detection of Adrenergic β3 receptor.
Western blot: 1:2,000-1:5,000. The antibody recognizes a band at ~44 kDa on mouse and rat brain lysate (40 μg).

Optimal working dilutions must be determined by end user.

Physical form

Liquid in PBS with 0.02% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer M Spaethling et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(1), 81-92 (2015-08-26)
Brown adipocytes (BAs) are specialized for adaptive thermogenesis and, upon sympathetic stimulation, activate mitochondrial uncoupling protein (UCP)-1 and oxidize fatty acids to generate heat. The capacity for brown adipose tissue (BAT) to protect against obesity and metabolic disease is recognized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service