Skip to Content
Merck
All Photos(1)

Key Documents

AB15467P

Sigma-Aldrich

Anti-ARG3.1 Antibody

Chemicon®, from rabbit

Synonym(s):

Growth factor ARC, Activity regulated cytoskeleton

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NOL3(8996)

General description

Arc/Arg3.1 is an immediate-early gene whose mRNA is rapidly transcribed and targeted to dendrites of neurons as they engage in information processing and storage. Arc/Arg3.1 is known to be required for durable forms of synaptic plasticity and learning. Arc/Arg3.1 has been shown to be expressed in neuronal cell bodies and dendrites in the brain regions including striatum and cortex hippocampus.

Specificity

ARG3.1 [Growth factor ARC, Activity regulated cytoskeleton]

Immunogen

Synthetic peptide corresponding to a region within the C-terminus of human ARG3.1 protein.

Application

Anti-ARG3.1 Antibody detects level of ARG3.1 & has been published & validated for use in IH.
Immunohistochemistry: 1:200-1:1,000

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Synapse & Synaptic Biology

Physical form

Format: Purified
Lyophilized. Reconstitute with 50 μL of sterile distilled water. Centrifuge to remove any insoluble material. Contains no preservative.

Storage and Stability

Maintain lyophilized material frozen at -20°C to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Glycerol (1:1, ACS or better grade) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service