Skip to Content
Merck
All Photos(1)

Key Documents

AB1542

Sigma-Aldrich

Anti-Tyrosine Hydroxylase Antibody

CHEMICON®, sheep polyclonal

Synonym(s):

Tyrosine 3-hydroxylase, Tyrosine 3-hydroxylase, tyrosine hydroxylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from sheep

biological source

sheep

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TH(7054)

General description

Tyrosine hydroxylase plays an important role in the physiology of adrenergic neurons. It is the first and rate-limiting enzyme involved in the biosynthesis of the catecholamines Dopamine and Norepinephrine from tyrosine. TH is, therefore, a useful marker for dopaminergic and noradrenergic neurons. The enzymatic activity of TH requires ferrous ions as cofactors and is believed to be regulated by phosphorylation. At least four isoforms of human TH have been identified which result from alternative splicing.

Specificity

Tyrosine Hydroxylase (TH, Tyrosine Monooxygenase). Cross-reacts with all mammalian forms tested to date and some non-mammalian forms.

Immunogen

Native tyrosine hydroxylase from rat pheochromocytoma (J. Biol. Chem.,1982, 257:9416-9423).

Application

Anti-Tyrosine Hydroxylase Antibody is an antibody against Tyrosine Hydroxylase for use in IH & WB.
Immunohistochemistry(peroxidase): A previous lot of this antibody was used at 1:1,000 dilution. The antibody gives specific labeling of noradrenergic axons in primate cerebral cortex (Brain Res., 1989, 500:313-324.).
Note: It is important that you use a secondary antibody that has significant reactivity to sheep IgG1 since the majority of the sheep immunoglobulin is IgG1.
Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Quality

Routinely evaluated by Western Blot on mouse brain lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected tyrosine hydroxylase on 10 μg of mouse brain lysates.

Target description

~60 kDa

Physical form

ImmunoAffinity Purified
Purified sheep polyclonal in buffer containing 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Stable for 6 months at -20 to -80°C in undiluted aliquots from date of receipt. Store in a NON-self-defrosting freezer.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Positive: brain (corpus striatum) and adrenal gland. Negative: liver.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective optogenetic activation of rostral ventrolateral medullary catecholaminergic neurons produces cardiorespiratory stimulation in conscious mice.
Abbott, SB; DePuy, SD; Nguyen, T; Coates, MB; Stornetta, RL; Guyenet, PG
The Journal of Neuroscience null
Transsynaptic activity-dependent regulation of axon branching and neurotrophin expression in vivo.
Calinescu, AA; Liu, T; Wang, MM; Borjigin, J
The Journal of Neuroscience null
Muriel T Davisson et al.
PloS one, 6(12), e29538-e29538 (2012-01-14)
Mutations in the gene encoding the immunoglobulin-superfamily member cell adhesion molecule contactin1 (CNTN1) cause lethal congenital myopathy in human patients and neurodevelopmental phenotypes in knockout mice. Whether the mutant mice provide an accurate model of the human disease is unclear;
Joel C Geerling et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 310(1), R41-R54 (2015-10-23)
The parabrachial nucleus is important for thermoregulation because it relays skin temperature information from the spinal cord to the hypothalamus. Prior work in rats localized thermosensory relay neurons to its lateral subdivision (LPB), but the genetic and neurochemical identity of
P R Brumovsky et al.
Neuroscience, 223, 77-91 (2012-08-04)
Previous studies in rat and mouse documented that a subpopulation of dorsal root ganglion (DRG) neurons innervating non-visceral tissues express tyrosine hydroxylase (TH). Here we studied whether or not mouse DRG neurons retrogradely traced with Fast Blue (FB) from colorectum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service