Skip to Content
Merck
All Photos(1)

Key Documents

AB15171

Sigma-Aldrich

Anti-Plasticity Related Gene 3 Antibody

Chemicon®, from rabbit

Synonym(s):

PRG-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

rat (immunogen homology), human (immunogen homology)

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

The previously assigned protein identifier Q5VX23 has been merged into Q8TBJ4. Full details can be found on the UniProt database.

Specificity

Plasticity related gene 3 (PRG-3). By Western blot the antibody recognizes a band at ~35 kDa in mouse brain lysate. An additional band at ~77 kDa may be seen depending on sample and antibody concentration used.

Immunogen

Synthetic peptide from human/rat/mouse PRG-3.

Application

Plasticity Related Gene 3 is an antibody targeting the Plasticity Related Gene 3 protein, validated for use in WB.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service