Skip to Content
Merck
All Photos(1)

Documents

AB10050

Sigma-Aldrich

Anti-phospho-ITCH (pT222) Antibody

Chemicon®, from rabbit

Synonym(s):

Itchy homolog E3 ubiquitin protein ligase, AIP4, NAPP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr222)

Gene Information

human ... ITCH(83737)

General description

ITCH is an E3 ubiquitin-protein ligase that accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. ITCH regulates the transcriptional activity of several transcription factors including the hematopoietic transcription factor NF-E2, and may play an important role in the regulation of immune response. ITCH down-regulates Epstein-Barr virus LMP2A activity in B cell signaling, and also interacts with NOTCH1, OCLN, JUN, and JUNB. ITCH has a wide tissue distribution and is localized within the nucleus and cytoplasm. Two ITCH splice variants have been described; an approximately 103 kDa form as well as a truncated form (~98 kDa) lacking 42 amino acids in the N-terminal half of the protein. ITCH contains a MAP Kinase binding site and its activity is modulated by phosphorylation at multiple threonine residues by JNK1.

Specificity

Human ITCH phosphorylated on threonine 222 (T222).

Immunogen

Synthetic peptide corresponding to a portion of human ITCH encompassing the phosphorylated Threonine at position 222 (PPPpTPRRP).

Application

Research Category
Protein Trafficking
Research Sub Category
Ubiquitin & Ubiquitin Metabolism
This Anti-phospho-ITCH (pT222) Antibody is validated for use in DB, WB for the detection of phospho-ITCH (pT222).
Western Blot: Recommended Dilution 1 - 5 μg/mL
Dot Blot

Physical form

Affinity purified immunoglobulin. Liquid in Tris-citrate/phosphate buffer, pH 7-8, containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C for up to 12 months from date of receipt

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Deng et al.
Journal of virology, 96(6), e0181121-e0181121 (2022-01-20)
We previously reported that hepatitis C virus (HCV) infection activates the reactive oxygen species (ROS)/c-Jun N-terminal kinase (JNK) signaling pathway. However, the roles of ROS/JNK activation in the HCV life cycle remain unclear. We sought to identify a novel role
Nikolaos Charlaftis et al.
The EMBO journal, 33(21), 2581-2596 (2014-09-28)
Unlike the other MAP3Ks, MEKK1 (encoded by Map3k1) contains a PHD motif. To understand the role of this motif, we have created a knockin mutant of mouse Map3k1 (Map3k1(m) (PHD)) with an inactive PHD motif. Map3k1(m) (PHD) ES cells demonstrate
Eutteum Jeong et al.
Nature communications, 15(1), 93-93 (2024-01-04)
Lysosomes have emerged as critical regulators of cellular homeostasis. Here we show that the lysosomal protein TMEM55B contributes to restore cellular homeostasis in response to oxidative stress by three different mechanisms: (1) TMEM55B mediates NEDD4-dependent PLEKHM1 ubiquitination, causing PLEKHM1 proteasomal
William J Sullivan et al.
Cell, 175(1), 117-132 (2018-09-11)
The metabolic state of a cell is influenced by cell-extrinsic factors, including nutrient availability and growth factor signaling. Here, we present extracellular matrix (ECM) remodeling as another fundamental node of cell-extrinsic metabolic regulation. Unbiased analysis of glycolytic drivers identified the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service