AAWG02500
MCE Membrane Filter, 0.8 μm Pore Size
MF-Millipore™, filter diam. 25 mm, hydrophilic, white, gridded
Synonym(s):
MF-Millipore™ Membrane Filter, 0.8 µm pore size, gridded
About This Item
Recommended Products
material
gridded
mixed cellulose esters (MCE) membrane
white filter
Quality Level
description
25 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, gridded, 100 discs
sterility
non-sterile
feature
hydrophilic
manufacturer/tradename
MF-Millipore™
Millipore
parameter
16 L/min-cm2 air flow rate
190 mL/min-cm2 water flow rate
75 °C max. temp.
diam.
25 mm
filter diam.
25 mm
thickness
150 μm
color
white
refractive index
n/D 1.51
matrix
MF-Millipore™
pore size
0.8 μm pore size
82 % porosity
bubble point
≥1.0 bar, air with water at 23 °C
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Application
- Air monitoring
- Particle monitoring
- Particle removal
- Bioassays
- Microplastics
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service