Skip to Content
Merck
All Photos(1)

Documents

960210

Supelco

Filling nozzle (tap)

self-closing, for use with 10-30L drums and barrels with thread G3/8

Synonym(s):

Filling nozzle horizontal supply

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.51

feature

self-closing

compatibility

for use with 10-30L drums and barrels with thread G3/8

shipped in

ambient

storage temp.

room temp

General description

Different environments and individual customer applications require special withdrawal options. Optimized nozzles are available for each special application, e.g. a seperate filling nozzle for individual length of tubing. This filling nozzle is a spare part to 9.67100.9090.
self-closing, for 10-30 L drums and barrels, with thread G3/8

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service