8590-OP
Sucrose solution
OmniPur®, 20% W/V
Synonym(s):
OmniPur® Sucrose Solution, 20% W/V
About This Item
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
haze-free colorless to slightly yellow
shipped in
ambient
General description
Molecular biology grade.
Specifications:
Clarity: To pass test
DNase: None detected
Protease: None detected
RNase: None detected
Sterility: To pass test
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Application
- Traditional Japanese medicine Kamikihito ameliorates sucrose preference, chronic inflammation, and obesity induced by a high fat diet in middle-aged mice.: Examines the effects of traditional Japanese medicine on the preference for sucrose solutions and its implications for dietary health, highlighting its potential for nutritional interventions (Maejima et al., 2024).
- Prenatal High-Sucrose Diet Induced Vascular Dysfunction of Renal Interlobar Arteries in the Offspring via PPARγ-RXRg-ROS/Akt Signaling.: Explores the developmental impacts of high-sucrose diets during pregnancy on offspring, particularly focusing on vascular health and potential intergenerational effects (Feng et al., 2024).
- Feeding Appropriate Nutrients during the Adult Stage to Promote the Growth and Development of Carposina sasakii Offspring.: Analyzes the effects of different nutrient solutions, including sucrose, on the growth and development of insect populations, with implications for biological research and pest management (Fu et al., 2024).
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service