Skip to Content
Merck
All Photos(1)

Key Documents

8.57005

Sigma-Aldrich

Capping Reagent B1 for DNA Synthesis

40% Acetic anhydride in MeCN (v/v), Novabiochem®

Synonym(s):

40% Acetic anhydride in MeCN (v/v) , Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

General description

Pre-mixed ready-to-use Capping Reagent
  • 857005.0500: Compatible with Cytiva ÄKTA DNA Synthesisers. Attaches directly to Instrument (GL-45 Neck Finish)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.9 °F

Flash Point(C)

9.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service