Skip to Content
Merck
All Photos(1)

Key Documents

8.55122

Sigma-Aldrich

NovaPEG Wang resin

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51171641
Pricing and availability is not currently available.

Quality Level

product line

NovaPEG
Novabiochem®

form

liquid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

shipped in

ambient

storage temp.

2-8°C

General description

An excellent support for the synthesis of peptide acids by Fmoc SPPS. The support has excellent swelling properties in a wide range of solvents, including water, MeCN, MeOH, DCM, DMF, THF, and toluene. In comparative studies this resin was found to give better results than polystyrene-based supports in the synthesis of hydrophobic peptides. The high hydrophilicity was also found to benefit on-resin immunoassays with one-bead-one-peptide libraries [1].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] F. Albericio, et al. Poster presented at 18th American Peptide Symposium, San Diego, June 2005.
[1] F. Garcia-Martin, et al. (2006) J. Comb. Chem., 8, 213.

Linkage

Replaces: 01-64-0474

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Protocols

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service