Skip to Content
Merck

Skip To

8.55005

NovaSyn®TGA resin (90 µm)

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
NACRES:
NA.22
UNSPSC Code:
12352203
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

product line

NovaSyn® TG, Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-30°C

General description

NovaSyn TGA is an excellent resin for both the continuous flow and batch Fmoc SPPS of peptide acids. It is derived from 90 μm NovaSyn TG amino resin by derivatization with the TFA labile 4-hydroxymethylphenoxyacetic acid linker.
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): white to pale yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.20 - 0.30 mmol/g
Swelling Volume (in DMF): lot specific result
The base resin is PEG-PS-copolymer (90µm) functionalized wit h 4-hydroxy-methylphenoxyacetic acid.

Other Notes

Replaces: 01-64-0040

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.550078.550098.55127
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

functional group

alcohol

functional group

amine

functional group

amine

functional group

amine

product line

NovaSyn® TG, Novabiochem®

product line

NovaSyn® TG, Novabiochem®

product line

NovaSyn® TG, Novabiochem®

product line

NovaSyn® TG, Novabiochem®

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-30°C


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Protocols

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Articles

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.





Global Trade Item Number

SKUGTIN
855005000504027269109301
855005000104027269109295
855005002504027269109318

Questions

Reviews

No rating value

Active Filters