Skip to Content
Merck
All Photos(1)

Documents

8.52419

Sigma-Aldrich

Fmoc-Cys(THP)-OH

Novabiochem®

Synonym(s):

Fmoc-Cys(THP)-OH, N-α-Fmoc-S-(tetrahydropyranyl)-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25NO5S
CAS Number:
Molecular Weight:
427.51
UNSPSC Code:
12352209

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

thiol

storage temp.

15-25°C

General description

Fmoc-Cys(Thp)-OH is a new building block for Fmoc SPPS, in which the sulfhydryl group is protected with the acid-cleavable tetrahydropyranyl (Thp) group [1]. The use of Fmoc-Cys(Thp)-OH has been shown to give superior results to the corresponding S-Trt, S-Dpm, S-Acm, and S-StBu derivatives.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Fmoc SPPS of Cysteine-Containing Peptides

Analysis Note

Colour (visual): white to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service