Skip to Content
Merck
All Photos(1)

Documents

8.52071

Sigma-Aldrich

Fmoc-Tyr(PO(OBzl)OH)-OH

≥95.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Tyr(PO(OBzl)OH)-OH, N-α-Fmoc-O-benzyl-L-phosphotyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H28NO8P
CAS Number:
Molecular Weight:
573.53
UNSPSC Code:
12352209
NACRES:
NA.22

product name

Fmoc-Tyr(PO(OBzl)OH)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥90.0% (acidimetric)
≥95% (TLC)
≥95.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

90-105 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

InChI

1S/C31H28NO8P/c33-30(34)29(18-21-14-16-23(17-15-21)40-41(36,37)39-19-22-8-2-1-3-9-22)32-31(35)38-20-28-26-12-6-4-10-24(26)25-11-5-7-13-27(25)28/h1-17,28-29H,18-20H2,(H,32,35)(H,33,34)(H,36,37)/t29-/m0/s1

InChI key

WNFKGEXQEWKECX-LJAQVGFWSA-N

General description

An excellent building block for the preparation of phosphotyrosine-containing peptides [1] by Fmoc SPPS. This derivative can be introduced using standard activation methods, such as PyBOP® and TBTU. Using this reagent, even peptides containing multiple phosphorylation sites have been prepared efficiently by standard Fmoc SPPS methods [1].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS


Literature references

[1] P. White & J. Beythien in ′Innovations & Perspectives in Solid Phase Synthesis and Combinatorial Libraries, 4th International Symposium′, Mayflower Scientific Ltd., Birmingham, 1996, pp. 557.

Linkage

Replaces: 04-12-1156

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 98.5 % (a/a)
Purity (TLC(CMA1)): ≥ 95 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 1.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.1 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

We provide an overview of our available reagents, together with recommendations and details of their use for synthesis of peptides containing post-translationally modified amino acids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service